Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CYP3A4 Antibodies

Bioss

CYP3A4 Polyclonal Antibody, Alexa Fluor™ 750

View all (29) CYP3A4 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CYP3A4 Polyclonal Antibody, Alexa Fluor™ 750

CYP3A4 Polyclonal Antibody, Alexa Fluor™ 750

Product Details

BS-1742R-A750

Applications
Tested Dilution
Publications

Western Blot (WB)

1:300-5,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

Immunohistochemistry (Frozen) (IHC (F))

1:50-1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

Flow Cytometry (Flow)

1:20-1:100
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic peptide derived from human CYP3A4
View immunogen

Conjugate

Alexa Fluor™ 750 Alexa Fluor™ 750 Alexa Fluor™ 750

Excitation/Emission Max

752/776 nm View spectra spectra

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

TBS, pH 7.4, with 1% BSA, 50% glycerol

Contains

0.03% ProClin 300

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. This enzyme is involved in the metabolism of approximately half the drugs in use today, including acetaminophen, codeine, cyclosporin A, diazepam and erythromycin. The enzyme also metabolizes some steroids and carcinogens. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Previously another CYP3A gene, CYP3A3, was thought to exist; however, it is now thought that this sequence represents a transcript variant of CYP3A4. Alternatively spliced transcript variants encoding different isoforms have been identified.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 1,4-cineole 2-exo-monooxygenase; 1,8-cineole 2-exo-monooxygenase; albendazole monooxygenase; Albendazole monooxygenase (sulfoxide-forming); Albendazole sulfoxidase; Cholesterol 25-hydroxylase; CYP; CYPIIIA3; CYPIIIA4; Cytochrome P450 3A3; Cytochrome P450 3A4; Cytochrome P450 HLp; Cytochrome P450 NF-25; cytochrome P450, family 3, subfamily A, polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; Cytochrome P450-PCN1; glucocorticoid-inducible P450; MGC126680; Nifedipine oxidase; P450; P450-III, steroid inducible; P450-PCN1; Quinine 3-monooxygenase; taurochenodeoxycholate 6-alpha-hydroxylase

View more View less

Gene Aliases: CP33; CP34; CYP3A; CYP3A3; CYP3A4; CYPIIIA3; CYPIIIA4; HLP; NF-25; P450C3; P450PCN1

View more View less

UniProt ID: (Human) P08684

View more View less

Entrez Gene ID: (Human) 1576

View more View less

Function(s)
monooxygenase activity steroid binding iron ion binding steroid hydroxylase activity oxidoreductase activity oxygen binding enzyme binding heme binding vitamin D3 25-hydroxylase activity taurochenodeoxycholate 6alpha-hydroxylase activity caffeine oxidase activity albendazole monooxygenase activity quinine 3-monooxygenase activity testosterone 6-beta-hydroxylase activity aromatase activity vitamin D 24-hydroxylase activity
Process(es)
lipid metabolic process steroid catabolic process xenobiotic metabolic process steroid metabolic process androgen metabolic process alkaloid catabolic process monoterpenoid metabolic process drug metabolic process calcitriol biosynthetic process from calciol vitamin D metabolic process drug catabolic process exogenous drug catabolic process heterocycle metabolic process oxidation-reduction process oxidative demethylation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0