Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CYP11B2 Antibodies

Invitrogen

CYP11B2 Polyclonal Antibody

View all (10) CYP11B2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CYP11B2 Polyclonal Antibody

CYP11B2 Polyclonal Antibody

Product Details

PA5-67694

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:3,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthesized peptide derived from human CYP11B2.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2691688

Product Specific Information

Predicted to react with Mouse and Rat samples.

Target Information

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the mitochondrial inner membrane and is involved in the conversion of progesterone to cortisol in the adrenal cortex. Mutations in this gene cause congenital adrenal hyperplasia due to 11-beta-hydroxylase deficiency. Transcript variants encoding different isoforms have been noted for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: ALDOS; Aldosterone synthase; Aldosterone-synthesizing enzyme; Corticosterone 18-monooxygenase, CYP11B2; CYPXIB2; CYPXIB3; Cytochrome P-450Aldo; Cytochrome P-450C18; Cytochrome P450 11B2, mitochondrial; cytochrome P450 11B3, mitochondrial; Cytochrome P450 subfamily XIB polypeptide 2 (aldosterone synthase); cytochrome P450, family 11, subfamily B, polypeptide 2; cytochrome P450, subfamily 11B, polypeptide 2; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 2; Cytochrome P450, subfamily XIB, polypeptide 2 (aldosterone synthase); cytochrome P450-Aldo-1; cytochrome P450-Aldo-2; Cytochrome P450C11; mitochondrial cytochrome P450, family 11, subfamily B, polypeptide 2; P-450C 18; P450-Aldo-1; P450-Aldo-2; P450C 18; steroid 11-beta-hydroxylase; Steroid 11-beta-hydroxylase, CYP11B2; steroid 11-beta-monooxygenase; steroid 11-beta/18-hydroxylase; Steroid 18-hydroxylase; steroid 18-hydroxylase, aldosterone synthase, P450C18, P450aldo; steroid-11-beta-hydroxylase

View more View less

Gene Aliases: ALDOS; Cp45as; CPN2; CYP11B; Cyp11b-2; CYP11B2; Cyp11b3; CYP11BL; CYPXIB2; P-450C18; P450aldo; P450C18; RNCP45AS

View more View less

UniProt ID: (Human) P19099, (Mouse) P15539

View more View less

Entrez Gene ID: (Human) 1585, (Mouse) 13072, (Rat) 24294

View more View less

Function(s)
steroid 11-beta-monooxygenase activity iron ion binding steroid hydroxylase activity oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen heme binding corticosterone 18-monooxygenase activity monooxygenase activity oxidoreductase activity oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen metal ion binding
Process(es)
regulation of systemic arterial blood pressure by circulatory renin-angiotensin regulation of blood volume by renal aldosterone steroid biosynthetic process C21-steroid hormone biosynthetic process glucocorticoid biosynthetic process regulation of blood pressure aldosterone biosynthetic process cortisol biosynthetic process drinking behavior positive regulation of blood pressure ion homeostasis oxidation-reduction process response to nutrient response to salt stress cellular response to nutrient response to insulin cellular response to hormone stimulus cellular response to potassium ion response to drug response to peptide hormone cellular response to potassium ion starvation potassium ion homeostasis sodium ion homeostasis cellular response to mechanical stimulus cellular response to peptide hormone stimulus renal water homeostasis mineralocorticoid biosynthetic process cholesterol metabolic process sterol metabolic process secondary metabolite biosynthetic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0