Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CtBP2 Antibodies

NeoBiotechnologies

CTBP2 (C-terminal binding protein 2) Monoclonal Antibody (PCRP-CTBP2-2D11)

View all (29) CtBP2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CTBP2 (C-terminal binding protein 2) Monoclonal Antibody (PCRP-CTBP2-2D11)

  • Antibody Testing Data (4)
CTBP2 (C-terminal binding protein 2) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
CTBP2 (C-terminal binding protein 2) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CTBP2 (C-terminal binding protein 2) Antibody (1488-MSM4-P1) in ICC/IF

Immunocytochemistry/Immunofluorescence analysis of MCF-7 cells using CTBP2 (C-terminal binding protein 2) Monoclonal Antibody (Product # 1488-MSM4-P1) followed by goat anti-mouse IgG-CF488 (green). Phalloidin (red).. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CTBP2 (C-terminal binding protein 2) Antibody in Immunocytochemistry (ICC/IF)
CTBP2 (C-terminal binding protein 2) Antibody in Flow Cytometry (Flow)
CTBP2 (C-terminal binding protein 2) Antibody in Peptide array (ARRAY)
CTBP2 (C-terminal binding protein 2) Antibody in SDS-PAGE (SDS-PAGE)
CTBP2 (C-terminal binding protein 2) Monoclonal Antibody (PCRP-CTBP2-2D11)

Product Details

1488-MSM4-P1

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

1-2 µg/mL
-

Flow Cytometry (Flow)

1-2 µg/10^6 cells
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

PCRP-CTBP2-2D11

Immunogen

Recombinant full-length human CTBP2 protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

This gene produces alternative transcripts encoding two distinct proteins. One protein is a transcriptional repressor, while the other isoform is a major component of specialized synapses known as synaptic ribbons. Both proteins contain a NAD+ binding domain similar to NAD+-dependent 2-hydroxyacid dehydrogenases. A portion of the 3' untranslated region was used to map this gene to chromosome 21q21. 3; however, it was noted that similar loci elsewhere in the genome are likely. Blast analysis shows that this gene is present on chromosome 10.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: C-terminal-binding protein 2; CtBP2; ribeye; RP11-59C5.2

View more View less

Gene Aliases: CTBP2

View more View less

UniProt ID: (Human) P56545

View more View less

Entrez Gene ID: (Human) 1488

View more View less

Function(s)
chromatin binding transcription coactivator activity transcription corepressor activity protein binding oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor protein homodimerization activity retinoic acid receptor binding NAD binding
Process(es)
negative regulation of transcription from RNA polymerase II promoter transcription, DNA-templated negative regulation of cell proliferation viral genome replication positive regulation of chromatin binding negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of retinoic acid receptor signaling pathway white fat cell differentiation oxidation-reduction process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-585975bbdc-jwj8f:80/100.66.129.203:80.
git-commit: a17be567db00e29ba041ba55be3678c43488a11a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0