Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CSK Antibodies

Proteintech

CSK Monoclonal Antibody (3B12C5), CoraLite® Plus 488

View all (31) CSK antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CSK Monoclonal Antibody (3B12C5), CoraLite® Plus 488

  • Antibody Testing Data (1)
CSK Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
CSK Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CSK Antibody (CL48866115100UL) in ICC/IF

Immunofluorescent analysis of (-20°C Ethanol) fixed A431 cells using CoraLite 488-conjugated CSK antibody (Product # CL48866115100UL), Clone: 3B12C5 ) at dilution of 1:100. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CSK Antibody in Immunocytochemistry (ICC/IF)
CSK Monoclonal Antibody (3B12C5), CoraLite® Plus 488

Product Details

CL48866115100UL

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

1:50-1:500
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

3B12C5

Conjugate

CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
  • Unconjugated
  • CoraLite 594
  • Request custom conjugation

Excitation/Emission Max

493/522 nm

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 50% glycerol, 0.5% BSA

Contains

0.05% ProClin 300

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles, store in dark

Shipping conditions

Wet ice

Target Information

CSK is a tyrosine kinase that has been shown to downregulate the activity of SRC oncoprotein through phosphorylation of its carboxy terminus. Since cell transformation by SRC is caused by various mechanisms that interfere with this phosphorylation, the CSK gene might function as an antioncogene. CSK plays an important role in the regulation of cell growth, differentiation, migration, and immune responses. CSK phosphorylates tyrosine residues located in the C-terminal tails of Src-family kinases including LCK, SRC, HCK, FYN, LYN, CSK, and YES1. CSK is expressed ubiquitously as a protein of approximately 50 KDa in cytosol, but a fraction distributes to lipid rafts due to interaction with the lipid raft-associated protein PAG (Csk-binding protein).

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: C-Src kinase; MGC117393; p50CSK; Protein-tyrosine kinase CYL; Protein-tyrosine kinase MPK-2; Tyrosine-protein kinase CSK

View more View less

Gene Aliases: AW212630; CSK; p50CSK

View more View less

UniProt ID: (Human) P41240, (Mouse) P41241

View more View less

Entrez Gene ID: (Human) 1445, (Mouse) 12988

View more View less

Function(s)
protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity receptor binding protein binding ATP binding protein C-terminus binding kinase activity protein phosphatase binding protein kinase A catalytic subunit binding identical protein binding metal ion binding proline-rich region binding
Process(es)
regulation of cytokine production adaptive immune response protein phosphorylation transmembrane receptor protein tyrosine kinase signaling pathway epidermal growth factor receptor signaling pathway central nervous system development brain development negative regulation of cell proliferation negative regulation of low-density lipoprotein particle clearance cell migration cell differentiation T cell costimulation negative regulation of interleukin-6 production negative regulation of kinase activity adherens junction organization peptidyl-tyrosine autophosphorylation regulation of cell proliferation negative regulation of Golgi to plasma membrane protein transport positive regulation of MAP kinase activity innate immune response negative regulation of bone resorption oligodendrocyte differentiation negative regulation of phagocytosis T cell receptor signaling pathway regulation of Fc receptor mediated stimulatory signaling pathway negative regulation of ERK1 and ERK2 cascade cellular response to peptide hormone stimulus
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0