Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • COX7A1 Antibodies

Invitrogen

COX7A1 Polyclonal Antibody

1 Published Figure
1 Reference
View all (4) COX7A1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite COX7A1 Polyclonal Antibody

  • Published Figures (1)
COX7A1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
COX7A1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

COX7A1 Antibody (PA5-67696) in WB

NPJ breast cancer 2024 - Fig. 5 HER2 patient proteomics. Stacked bar chart representing the number of unique proteins detected by the number of samples ( a n = 10 biopsies, n = 1555 unique proteins). Differential expression analysis of detected proteins, represented as a volcano plot with significantly differentially expressed proteins (DEPs) identified in black ( b ). Heatmap of DEPs, with the ten columns representing the ten samp... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

COX7A1 Antibody in Western Blot (WB)
COX7A1 Polyclonal Antibody

Product Details

PA5-67696

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:3,000
View 1 publication 1 publication
Product Specifications

Species Reactivity

Human

Published species

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthesized peptide derived from human COX7A1.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2691462

Target Information

This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: Cytochrome c oxidase subunit 7A1, mitochondrial; cytochrome c oxidase subunit VIIa heart/muscle isoform; cytochrome c oxidase subunit VIIa polypeptide 1 (muscle); Cytochrome c oxidase subunit VIIa-H; Cytochrome c oxidase subunit VIIa-heart; Cytochrome c oxidase subunit VIIa-M; Cytochrome c oxidase subunit VIIa-muscle

View more View less

Gene Aliases: COX7A; COX7A1; COX7AH; COX7AM

View more View less

UniProt ID: (Human) P24310

View more View less

Entrez Gene ID: (Human) 1346

View more View less

Function(s)
cytochrome-c oxidase activity
Process(es)
generation of precursor metabolites and energy hydrogen ion transmembrane transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0