Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • COX5A Antibodies

Invitrogen

COX5A Polyclonal Antibody

View all (14) COX5A antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite COX5A Polyclonal Antibody

  • Antibody Testing Data (5)
COX5A Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
COX5A Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

COX5A Antibody (PA5-121003) in ICC/IF

Immunofluorescence analysis of COX5A in U2OS cells. Samples were incubated with COX5A Polyclonal antibody (Product # PA5-121003). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

COX5A Antibody in Immunocytochemistry (ICC/IF)
COX5A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
COX5A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
COX5A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
COX5A Antibody in Western Blot (WB)
COX5A Polyclonal Antibody

Product Details

PA5-121003

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:100
-

Immunocytochemistry (ICC/IF)

1:10-1:100
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human COX5A (NP_004246.2).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

4.35 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2914575

Product Specific Information

Positive test controls include: HeLa, HepG2, mouse skeletal muscle, mouse testis, mouse heart, mouse brain, rat heart, rat kidney, Jurkat. The target is usually found in the following locations: Mitochondrion inner membrane.

Immunogen sequence: MLGAALRRCA VAATTRADPR GLLHSARTPG PAVAIQSVRC YSHGSQETDE EFDARWVTYF NKPDIDAWEL RKGINTLVTY DMVPEPKIID AALRACRRLN DFASTVRILE VVKDKAGPHK EIYPYVIQEL RPTLNELGIS TPEELGLDKV

Target Information

Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The mitochondrially-encoded subunits perform the electron transfer of proton pumping activities. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. This gene encodes the nuclear-encoded subunit Va of the human mitochondrial respiratory chain enzyme. A pseudogene COX5AP1 has been found in chromosome 14q22.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Cytochrome c oxidase polypeptide Va; cytochrome c oxidase polypeptide, mitochondrial; Cytochrome c oxidase subunit 5A, mitochondrial; cytochrome c oxidase subunit Va; mitochondrial cytochrome c oxidase subunit Va

View more View less

Gene Aliases: AA959768; CcOX; COX; COX-VA; COX5A; VA

View more View less

UniProt ID: (Human) P20674, (Rat) P11240, (Mouse) P12787

View more View less

Entrez Gene ID: (Human) 9377, (Rat) 252934, (Mouse) 12858

View more View less

Function(s)
cytochrome-c oxidase activity protein binding electron carrier activity metal ion binding
Process(es)
mitochondrial electron transport, cytochrome c to oxygen hydrogen ion transmembrane transport oxidation-reduction process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0