Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • COBRA1 Antibodies

Abnova

COBRA1 Polyclonal Antibody, MaxPab™

View all (13) COBRA1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite COBRA1 Polyclonal Antibody, MaxPab™

  • Antibody Testing Data (2)
COBRA1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
COBRA1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

COBRA1 Antibody (H00025920-D01P) in WB

Western Blot analysis of COBRA1 expression in transfected 293T cell line (H00025920-T02) by COBRA1 MaxPab polyclonal antibody.

Lane 1: COBRA1 transfected lysate(53.90 KDa).
Lane 2: Non-transfected lysate.
{{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

COBRA1 Antibody in Western Blot (WB)
COBRA1 Antibody in Western Blot (WB)

Product Details

H00025920-D01P

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

COBRA1 (AAH11892.1, 1 a.a. approximately 475 a.a) full-length human protein.

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: MPSLQPVVMC VMKHLPKVPE KKLKLVMADK ELYRACAVEV KRQIWQDNQA LFGDEVSPLL KQYILEKESA LFSTELSVLH NFFSPSPKTR RQGEVVQRLT RMVGKNVKLY DMVLQFLRTL FLRTRNVHYC TLRAELLMSL HDLDVGEICT VDPCHKFTWC LDACIRERFV DSKRARELQG FLDGVKKGQE QVLGDLSMIL CDPFAINTLA LSTVRHLQEL VGQETLPRDS PDLLLLLRLL ALGQGAWDMI DSQVFKEPKM EVELITRFLP MLMSFLVDDY TFNVDQKLPA EEKAPVSYPN TLPESFTKFL QEQRMACEVG LYYVLHITKQ RNKNALLRLL PGLVETFGDL AFGDIFLHLL TGNLALLADE FALEDFCSSL FDGFFLTASP RKENVHRHAL RLLIHLHPRV APSKLEALQK ALEPTGQSGE AVKELYSQLG EKLEQLDHRK PSPAQAAETP ALELPLPSVP APAPL

Target Information

NELFB is a subunit of negative elongation factor (NELF), which also includes NELFA (WHSC2), either NELFC or NELFD (TH1L), and NELFE (RDBP), a heterodimer of SPT4 (SUPT4H1), to cause transcriptional pausing of RNA polymerase II (Narita et al., 2003).

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Cofactor of BRCA1; DKFZp586B0519; Negative elongation factor B; negative elongation factor protein B; NELF-B; RP13-122B23.3

View more View less

Gene Aliases: COBRA1; KIAA1182; NELF-B; NELFB

View more View less

UniProt ID: (Human) Q8WX92

View more View less

Entrez Gene ID: (Human) 25920

View more View less

Function(s)
protein binding
Process(es)
transcription from RNA polymerase II promoter transcription elongation from RNA polymerase II promoter negative regulation of transcription elongation from RNA polymerase II promoter positive regulation of viral transcription
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0