Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CNOT6 Antibodies

Proteintech

CNOT6 Polyclonal Antibody

View all (5) CNOT6 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CNOT6 Polyclonal Antibody

  • Antibody Testing Data (1)
CNOT6 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
CNOT6 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CNOT6 Antibody (17935-1-AP) in WB

Jurkat cells were subjected to SDS PAGE followed by western blot with 17935-1-AP (CNOT6 Antibody) at dilution of 1:300 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CNOT6 Antibody in Western Blot (WB)
CNOT6 Polyclonal Antibody

Product Details

17935-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:200-1:1,000
-

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Purification

Antigen affinity chromatography

Storage buffer

PBS with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Immunogen sequence: MCIMTISSEG ELELVGLMRL RKQLFLRSAL CNHRSRELYG SGRGGALTHV VFVNGGCHLF IDAGTRVHLL DAKGLSFTQN VFICICYCLQ YI

Target Information

Poly(A) nuclease with 3'-5' RNase activity. Catalytic component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Involved in mRNA decay mediated by the major-protein-coding determinant of instability (mCRD) of the FOS gene in the cytoplasm. In the presence of ZNF335, enhances ligand-dependent transcriptional activity of nuclear hormone receptors, including RARA. The increase of ligand-dependent ESR1-mediated transcription is much smaller, if any. Mediates cell proliferation and cell survival and prevents cellular senescence.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: carbon catabolite repression 4 protein; Carbon catabolite repressor protein 4 homolog; CCR4 carbon catabolite repression 4-like; CCR4-NOT transcription complex subunit 6; CCR4-NOT transcription complex, subunit 6; Cytoplasmic deadenylase

View more View less

Gene Aliases: CCR4; CCR4a; CNOT6; KIAA1194

View more View less

UniProt ID: (Human) Q9ULM6, (Mouse) Q8K3P5

View more View less

Entrez Gene ID: (Human) 57472, (Mouse) 104625

View more View less

Function(s)
RNA binding exoribonuclease activity poly(A)-specific ribonuclease activity protein binding metal ion binding nuclease activity exonuclease activity hydrolase activity
Process(es)
transcription, DNA-templated regulation of transcription, DNA-templated regulation of translation positive regulation of cell proliferation positive regulation of cytoplasmic mRNA processing body assembly gene silencing by RNA exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay nuclear-transcribed mRNA catabolic process, no-go decay RNA phosphodiester bond hydrolysis, exonucleolytic positive regulation of ligand-dependent nuclear receptor transcription coactivator activity nuclear-transcribed mRNA poly(A) tail shortening DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest gene silencing by miRNA
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0