Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CLIC5 Antibodies

Invitrogen

CLIC5 Monoclonal Antibody (CLIC5-02)

View all (7) CLIC5 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CLIC5 Monoclonal Antibody (CLIC5-02)

  • Antibody Testing Data (2)
CLIC5 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
CLIC5 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CLIC5 Antibody (MA5-28624) in WB

Western blotting analysis of CLIC5a in HEK293-CLIC5a transfectants (A) and HEK293 cells (B) using mouse monoclonal antibody (clone CLIC5-02) (Product # MA5-28624). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CLIC5 Antibody in Western Blot (WB)
CLIC5 Antibody in Flow Cytometry (Flow)
CLIC5 Monoclonal Antibody (CLIC5-02)

Product Details

MA5-28624

Applications
Tested Dilution
Publications

Western Blot (WB)

Assay-dependent
-

Flow Cytometry (Flow)

1-4 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b, kappa

Class

Monoclonal

Type

Antibody

Clone

CLIC5-02

Immunogen

Human CLIC5a (amino acids 160-173)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated
  • Alexa Fluor 647 
  • PE 
  • Request custom conjugation

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

15mM sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2745583

Product Specific Information

This antibody recognizes CLIC5a, a 32 kDa intracellular protein which associates with proteins of actin complexes. Cross reactivity with CLIC5b was not determined.

Target Information

Clic5 encodes a protein that is required for normal hearing and is necessary for the formation of stereocilia in the inner ear and normal development of the organ of Corti. Clic5 can insert into membranes and form poorly selective ion channels that may also transport chloride ions. Clic5 may play a role in the regulation of transepithelial ion absorption and secretion, and is required for the development and/or maintenance of the proper glomerular endothelial cell and podocyte architecture. The Clic5 gene encodes a member of the chloride intracellular channel (CLIC) family of chloride ion channels. The protein encoded by Clic5 associates with actin-based cytoskeletal structures and may also play a role in multiple processes including hair cell stereocilia formation, myoblast proliferation and glomerular podocyte and endothelial cell maintenance. Alternatively spliced transcript variants encoding multiple isoforms have been observed for the Clic5 gene. Diseases associated with CLIC5 include Deafness, Autosomal Recessive 103 and Autosomal Recessive Non-Syndromic Sensorineural Deafness Type Dfnb.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Chloride intracellular channel protein 5; Glutaredoxin-like oxidoreductase CLIC5

View more View less

Gene Aliases: CLIC5; DFNB102; DFNB103; MST130; MSTP130

View more View less

UniProt ID: (Human) Q9NZA1

View more View less

Entrez Gene ID: (Human) 53405

View more View less

Function(s)
glutathione transferase activity voltage-gated ion channel activity chloride channel activity protein binding
Process(es)
diet induced thermogenesis glutathione metabolic process transport chloride transport female pregnancy sensory perception of sound protein localization regulation of ion transmembrane transport neuromuscular process controlling balance auditory receptor cell stereocilium organization chloride transmembrane transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0