Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CLEC9A Antibodies

NeoBiotechnologies

CLEC9A/DNGR1 (Dendritic Cell Marker) Monoclonal Antibody (8F9)

View all (24) CLEC9A antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CLEC9A/DNGR1 (Dendritic Cell Marker) Monoclonal Antibody (8F9)

  • Antibody Testing Data (1)
CLEC9A/DNGR1 (Dendritic Cell Marker) Antibody in SDS-PAGE (SDS-PAGE)
Group 53 Created with Sketch.
CLEC9A/DNGR1 (Dendritic Cell Marker) Antibody in SDS-PAGE (SDS-PAGE)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CLEC9A/DNGR1 (Dendritic Cell Marker) Antibody (283420-MSM2-P1ABX) in SDS-PAGE

SDS-PAGE analysis Purified CLEC9A Mouse Monoclonal Antibody (8F9). Confirmation of Integrity and Purity of Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CLEC9A/DNGR1 (Dendritic Cell Marker) Antibody in SDS-PAGE (SDS-PAGE)

Product Details

283420-MSM2-P1ABX

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

1-2 µg/mL
-

Flow Cytometry (Flow)

1-2 µg/10^6 cells
-

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a, kappa

Class

Monoclonal

Type

Antibody

Clone

8F9

Immunogen

RBL-2H3 cells expressing human DNGR-1 fused to an HA epitope

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

CLEC9A is a group V C-type lectin-like receptor (CTLR) that functions as an activation receptor and is expressed on myeloid lineage cells.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: C-type lectin domain family 9 member A; C-type lectin domain family 9, member A; CD370; HEEE9341

View more View less

Gene Aliases: CD370; CLEC9A; DNGR-1; DNGR1; UNQ9341; UNQ9341/PRO34046

View more View less

UniProt ID: (Human) Q6UXN8

View more View less

Entrez Gene ID: (Human) 283420

View more View less

Function(s)
carbohydrate binding
Process(es)
receptor-mediated endocytosis positive regulation of cytokine secretion
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-585975bbdc-x5cgg:80/100.66.128.25:80.
git-commit: a17be567db00e29ba041ba55be3678c43488a11a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0