Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QPAVQQAPT PAAPQPLQMG PCAYEIRQFL DCSTTQSDLS LCEGFSEALK QCKYYHGLSS LP (82-142 aa encoded by BC065232)
CHCHD10 encodes a mitochondrial protein that is enriched at cristae junctions in the intermembrane space. It may play a role in cristae morphology maintenance or oxidative phosphorylation. Mutations in this gene cause frontotemporal dementia and/or amyotrophic lateral sclerosis-2. Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7 and 19.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil-helix-coiled-coil-helix domain-containing protein 10, mitochondrial; Nur77 downstream gene 2; Nur77 downstream protein 2; Protein N27C7-4
Gene Aliases: 1620401E04Rik; AI267078; C22orf16; CHCHD10; FTDALS2; IMMD; N27C7-4; Ndg2; SMAJ
UniProt ID: (Human) Q8WYQ3
Entrez Gene ID: (Human) 400916, (Mouse) 103172
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support