Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • C/EBP epsilon Antibodies

Proteintech

CEBPE Polyclonal Antibody

1 Reference
View all (13) C/EBP epsilon antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CEBPE Polyclonal Antibody

  • Antibody Testing Data (3)
CEBPE Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
CEBPE Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CEBPE Antibody (14271-1-AP) in ICC/IF

Immunofluorescent analysis of ( 4% PFA) fixed HepG2 cells using 14271-1-AP (CEBPE antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CEBPE Antibody in Immunocytochemistry (ICC/IF)
CEBPE Antibody in Western Blot (WB)
CEBPE Antibody in Western Blot (WB)
CEBPE Polyclonal Antibody

Product Details

14271-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:200-1:1,000
View 1 publication 1 publication

Immunocytochemistry (ICC/IF)

1:50-1:500
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

CEBPE Fusion Protein Ag5589 (1-281 aa encoded by BC035797)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.23 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: MSHGTYYEC EPRGGQQPLE FSGGRAGPGE LGDMCEHEAS IDLSAYIESG EEQLLSDLFA VKPAPEARGL KGPGTPAFPH YLPPDPRPFA YPPHTFGPDR KALGPGIYSS PGSYDPRAVA VKEEPRGPEG SRAASRGSYN PLQYQVAHCG QTAMHLPPTL AAPGQPLRVL KAPLATAAPP CSPLLKAPSP AGPLHKGKKA VNKDSLEYRL RRERNNIAVR KSRDKAKRRI LETQQKVLEY MAENERLRSR VEQLTQELDT LRNLFRQIPE AANLIKGVGG CS (1-281 aa encoded by BC035797)

Target Information

C/EBP alpha regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. C/EBP alpha uses a bipartite structural motif to bind DNA. Two protein chains dimerize through a set of amphipathic alpha helices termed the leucine zipper. Highly basic polypeptide regions emerge from the zipper to form a linked set of DNA contact surfaces. C/EBP alpha appears to function exclusively in terminally-differentiated, growth-arrested cells. Additional family members include C/EBP beta, C/EBP gamma, C/EBP delta and C/EBP epsilon, all of which exhibit similar DNA-binding specificities and affinities to C/EBP alpha. Furthermore, C/EBP beta and C/EBP delta readily form heterodimers both with each other as well as with C/EBP alpha.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: C EBP; C/EBP epsilon; C/EBP-related protein 1; C/EBPepsilon; CCAAT/enhancer binding protein (C/EBP), epsilon; CCAAT/enhancer binding protein , epsilon; CCAAT/enhancer binding protein epsilon; CCAAT/enhancer-binding protein epsilon

View more View less

Gene Aliases: C/EBP-epsilon; C/EBPe; C/EPBe; CEBPE; CRP1; Gm294

View more View less

UniProt ID: (Human) Q15744, (Rat) P56261, (Mouse) Q6PZD9

View more View less

Entrez Gene ID: (Human) 1053, (Rat) 25410, (Mouse) 110794

View more View less

Function(s)
RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding protein homodimerization activity protein heterodimerization activity DNA binding transcription factor activity, sequence-specific DNA binding sequence-specific DNA binding
Process(es)
transcription, DNA-templated phagocytosis positive regulation of gene expression myeloid cell differentiation macrophage differentiation cytokine biosynthetic process defense response to bacterium positive regulation of transcription from RNA polymerase II promoter cellular response to lipopolysaccharide transcription from RNA polymerase II promoter defense response regulation of transcription, DNA-templated
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0