Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD66c (CEACAM6) Antibodies

Invitrogen

CEACAM6 Polyclonal Antibody

View all (27) CD66c (CEACAM6) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CEACAM6 Polyclonal Antibody

  • Antibody Testing Data (1)
CEACAM6 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
CEACAM6 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CEACAM6 Antibody (PA5-97055) in WB

Western blot analysis of CEACAM6 in A-549 cells. Samples were incubated with CEACAM6 Polyclonal antibody (Product # PA5-97055) using a dilution of 1:2,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 90s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CEACAM6 Antibody in Western Blot (WB)
CEACAM6 Polyclonal Antibody

Product Details

PA5-97055

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:5,000
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 35-320 of human CEACAM6 (NP_0024744)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1.37 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

RRID

AB_2808857

Product Specific Information

Positive Samples: A-549, Raji, HeLa, Mouse lung; Cellular Location: Cell membrane, GPI-anchor, Lipid-anchor

Immunogen sequence: KLTIESTPFN VAEGKEVLLL AHNLPQNRIG YSWYKGERVD GNSLIVGYVI GTQQATPGPA YSGRETIYPN ASLLIQNVTQ NDTGFYTLQV IKSDLVNEEA TGQFHVYPEL PKPSISSNNS NPVEDKDAVA FTCEPEVQNT TYLWWVNGQS LPVSPRLQLS NGNMTLTLLS VKRNDAGSYE CEIQNPASAN RSDPVTLNVL YGPDVPTISP SKANYRPGEN LNLSCHAASN PPAQYSWFIN GTFQQSTQEL FIPNITVNNS GSYMCQAHNS ATGLNRTTVT MITVSG

Target Information

CEA Cell Adhesion Molecule 6 (CEACAM6) belongs to the carcinoembryonic antigen (CEA) family whose members are glycosyl phosphatidyl inositol (GPI) anchored cell surface glycoproteins. Members of this family play a role in cell adhesion and are widely used as tumor markers in serum immunoassay determinations of carcinoma. CEACAM6 affects the sensitivity of tumor cells to adenovirus infection. The acts as a receptor for adherent-invasive E. coli adhesion to the surface of ileal epithelial cells in patients with Crohn's disease.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: BGP; BGP-1; Carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); CD66a; CD66c; CD66d; CD66e; CEA; CEA cell adhesion molecule 6; CEACAM-1; CEACAM-3; CEACAM-5; CEACAM-6; Cell adhesion molecule CEACAM6; CGM1; Cluster of Differentiation 66c; Non-specific crossreacting antigen; Normal cross-reacting antigen; pan CEACAM

View more View less

Gene Aliases: CD66c; CEACAM6; CEAL; NCA

View more View less

UniProt ID: (Human) P40199

View more View less

Entrez Gene ID: (Human) 4680

View more View less

Function(s)
protein binding
Process(es)
signal transduction cell-cell signaling leukocyte migration
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0