Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD90 (Thy1) Antibodies

Invitrogen

CD90 (Thy-1) Monoclonal Antibody (eBio5E10 (5E10)), PerCP-eFluor™ 710, eBioscience™

41 Published Figures
37 References
View all (73) CD90 (Thy1) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD90 (Thy-1) Monoclonal Antibody (eBio5E10 (5E10)), PerCP-eFluor™ 710, eBioscience™

  • Antibody Testing Data (1)
  • Published Figures (41)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 42

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD90 (Thy-1) Antibody (46-0909-42) in Flow

Staining of the HEL cell line with Mouse IgG1 K Isotype Control PerCP-eFluor® 710 (Product # 46-4714-82) (blue histogram) or Anti-Human CD90 (Thy-1) PerCP-eFluor® 710 (purple histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)
CD90 (Thy-1) Antibody in Flow Cytometry (Flow)

Product Details

46-0909-42

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

5 µL (0.125 µg)/test
View 37 publications 37 publications
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Recommended Isotype Control

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PerCP-eFluor™ 710, eBioscience™

Class

Monoclonal

Type

Antibody

Clone

eBio5E10 (5E10)

Conjugate

PerCP-eFluor™ 710 PerCP-eFluor™ 710 PerCP-eFluor™ 710
  • Unconjugated
  • Alexa Fluor 700 
  • APC 
  • APC-eFluor 780
  • Biotin 
  • Brilliant UV 395
  • Brilliant UV 563
  • Brilliant Violet 480
  • Brilliant Violet 650
  • Brilliant Violet 786
  • FITC
  • PE 
  • PE-Cyanine5
  • PE-Cyanine7
  • PerCP-Cyanine5.5
  • Super Bright 436
  • Request custom conjugation

Excitation/Emission Max

482/708 nm View spectra spectra

Form

Liquid

Concentration

5 µL/Test

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2, with 0.2% BSA

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_10671410

Product Specific Information

Description: The eBio5E10 monoclonal antibody reacts with human CD90, also known as Thy-1 (thymus cell antigen-1). CD90 is a 25-35 kDa receptor expressed on thymocytes, CD34+ prothymocytes, hematopoietic stem cells, neurons, a small subset of human fetal liver cells, cord blood cells, and bone marrow cells. CD90 is expressed on a subset of immature, CD34+ cells and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is enriched for cells capable of long-term culture. CD90 is involved in regulation of adhesion and signal transduction by T cells.

Applications Reported: This eBio5E10 (5E10) antibody has been reported for use in flow cytometric analysis.

Applications Tested: This eBio5E10 (5E10) antibody has been pre-titrated and tested by flow cytometric analysis of the HEL cell line. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.

Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.

Filtration: 0.2 µm post-manufacturing filtered.

Target Information

CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

Bioinformatics

Protein Aliases: CD90; CDw90; FLJ33325; Thy-1 antigen; Thy-1 membrane glycoprotein; Thy-1 T-cell antigen

View more View less

Gene Aliases: CD90; CDw90; THY1

View more View less

UniProt ID: (Human) P04216

View more View less

Entrez Gene ID: (Human) 7070

View more View less

Function(s)
GTPase activator activity integrin binding protein binding protein kinase binding GPI anchor binding
Process(es)
angiogenesis negative regulation of protein kinase activity cytoskeleton organization single organismal cell-cell adhesion negative regulation of cell migration positive regulation of GTPase activity retinal cone cell development protein autophosphorylation focal adhesion assembly negative regulation of axonogenesis T cell receptor signaling pathway negative regulation of T cell receptor signaling pathway positive regulation of T cell activation positive regulation of release of sequestered calcium ion into cytosol
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0