Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD8 Antibodies

Invitrogen

CD8a Monoclonal Antibody (53-6.7), Brilliant Ultra Violet™ 615, eBioscience™

View all (529) CD8 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD8a Monoclonal Antibody (53-6.7), Brilliant Ultra Violet™ 615, eBioscience™

  • Antibody Testing Data (1)
CD8a Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD8a Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD8a Antibody (366-0081-82) in Flow

C57BL/6 mouse splenocytes were stained with CD4 Monoclonal Antibody, FITC (Product # 11-0042-82) and 1.0 µg of Rat IgG2a kappa Isotype Control, Brilliant Ultra Violet 615 (Product # 366-4321-81) (left) or 1.0 µg of CD8a Monoclonal Antibody, Brilliant Ultra Violet 615 (right). Viable cells in the lymphocyte gate were used for analysis, as determined by 7-AAD (Product # 00-6993-50). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD8a Antibody in Flow Cytometry (Flow)

Product Details

366-0081-82

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

1.0 µg/test
-
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Rat / IgG2a, kappa

Recommended Isotype Control

Rat IgG2a kappa Isotype Control (eBR2a), Brilliant Ultra Violet™ 615, eBioscience™

Class

Monoclonal

Type

Antibody

Clone

53-6.7

Conjugate

Brilliant Ultra Violet™ 615 Brilliant Ultra Violet™ 615 Brilliant Ultra Violet™ 615
  • Unconjugated
  • Alexa Fluor 488 
  • Alexa Fluor 532 
  • Alexa Fluor 561
  • Alexa Fluor 660 
  • Alexa Fluor 700 
  • APC 
  • APC-eFluor 780
  • Biotin 
  • Brilliant UV 395
  • Brilliant UV 496
  • Brilliant UV 563
  • Brilliant UV 661
  • Brilliant UV 737
  • Brilliant UV 805
  • Brilliant Violet 421
  • Brilliant Violet 480
  • Brilliant Violet 605
  • Brilliant Violet 650
  • Brilliant Violet 711
  • Brilliant Violet 786
  • eFluor 450
  • eFluor 506
  • eFluor 615
  • eFluor 660
  • FITC
  • Functional Grade
  • NovaFluor Blue 510
  • NovaFluor Blue 585
  • NovaFluor Blue 610-70S
  • NovaFluor Blue 660-120S
  • NovaFluor Blue 725
  • NovaFluor Blue 760
  • NovaFluor Blue 800
  • NovaFluor Red 710
  • NovaFluor UV 765
  • NovaFluor Violet 690
  • NovaFluor Violet 745
  • NovaFluor Violet 800
  • NovaFluor Yellow 590
  • NovaFluor Yellow 610
  • NovaFluor Yellow 660
  • NovaFluor Yellow 690
  • NovaFluor Yellow 700
  • NovaFluor Yellow 730
  • NovaFluor Yellow 810
  • PE 
  • PE-Cyanine5
  • PE-Cyanine5.5
  • PE-Cyanine7
  • PE-eFluor 610
  • PerCP-Cyanine5.5
  • PerCP-eFluor 710
  • Super Bright 436
  • Super Bright 600
  • Super Bright 645
  • Super Bright 702
  • Super Bright 780
  • Request custom conjugation

Excitation/Emission Max

350/607 nm View spectra spectra

Form

Liquid

Concentration

0.2 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2, with BSA

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2920984

Product Specific Information

Description
The 53-6.7 monoclonal antibody reacts with the mouse CD8a molecule.

Applications Tested
This 53-6.7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 1.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Blocking Buffers
When using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) or Brilliant Stain Buffer (Product # 00-4409-75) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer or Brilliant Stain Buffer for more information.

Light sensitivity
This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

Fixation
• Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
• Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

Excitation: 350 nm; Emission: 615 nm; Laser: Ultraviolet Laser.

BRILLIANT ULTRA VIOLET™ is a trademark or registered trademark of Becton, Dickinson and Company or its affiliates, and is used under license. Powered by Sirigen™.

Target Information

CD8, also known as cluster of differentiation 8, is a type I transmembrane glycoprotein of the immunoglobulin family that plays a crucial role in T cell differentiation, activation, and signal transduction. It is expressed as either a heterodimer (CD8 alpha beta) or a homodimer (CD8 alpha alpha). The CD8 alpha beta form is predominantly found on the majority of thymocytes and a subpopulation of mature alpha beta TCR T cells, while the CD8 alpha alpha form is expressed on gamma delta TCR T cells, a subset of intestinal intraepithelial lymphocytes (IELs), and dendritic cells. CD8 functions as a co-receptor for major histocompatibility complex class I (MHC-I) molecules, working alongside the T cell receptor (TCR). The CD8 alpha chain is essential for binding to MHC-I. CD8 is also expressed on a subset of T cells, NK cells, monocytes, and dendritic cells as disulfide-linked homodimers of CD8 alpha. Upon ligation of MHC-I/peptide complexes presented by antigen-presenting cells (APCs), CD8 recruits lymphocyte-specific protein tyrosine kinase (Lck), leading to lymphokine production, increased motility, and activation of cytotoxic T lymphocytes (CTLs). Activated CTLs are vital for clearing pathogens and tumor cells. The differentiation of naive CD8+ T cells into CTLs is strongly enhanced by cytokines such as IL-2, IL-12, and TGF-beta1. Through its interactions with MHC-I and association with protein tyrosine kinase p56lck, CD8 plays a significant role in T cell development and the activation of mature T cells.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD8a; CD8alpha; CD8b; CD8beta; fCD8; Leu-2; leu-2a; Lymphocyte antigen 3; Lyt-2.1 lymphocyte differentiation antigen (AA at 100); T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-2; T-cell surface glycoprotein Lyt-3

View more View less

Gene Aliases: BB154331; Cd8a; Cd8b; Cd8b1; Ly-2; Ly-3; Ly-35; Ly-B; Ly-C; Lyt-2; Lyt-3; Lyt2; Lyt3

View more View less

UniProt ID: (Mouse) P01731, (Mouse) P10300

View more View less

Entrez Gene ID: (Mouse) 12525, (Mouse) 12526

View more View less

Function(s)
protein binding protein kinase binding protein homodimerization activity
Process(es)
immune system process T cell mediated immunity cell surface receptor signaling pathway T cell activation cytotoxic T cell differentiation positive regulation of calcium-mediated signaling defense response to virus
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0