Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD79a Antibodies

NeoBiotechnologies

CD79a (B-Cell Marker) Monoclonal Antibody (HM57)

View all (83) CD79a antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD79a (B-Cell Marker) Monoclonal Antibody (HM57)

CD79a (B-Cell Marker) Monoclonal Antibody (HM57)
  • Antibody Testing Data (5)
CD79a (B-Cell Marker) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
CD79a (B-Cell Marker) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD79a (B-Cell Marker) Antibody (973-MSM7-P0) in ICC/IF

Immunofluorescence analysis of PFA-fixed Raji cells labeling CD79a with CD79a Monoclonal Antibody (HM57) followed by Goat anti-Mouse IgG-CF488 (Green). The nuclear counterstain is RedDot (Red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD79a (B-Cell Marker) Antibody in Immunocytochemistry (ICC/IF)
CD79a (B-Cell Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CD79a (B-Cell Marker) Antibody in Western Blot (WB)
CD79a (B-Cell Marker) Antibody in Flow Cytometry (Flow)
CD79a (B-Cell Marker) Antibody in SDS-PAGE (SDS-PAGE)

Product Details

973-MSM7-P0

Applications
Tested Dilution
Publications

Western Blot (WB)

1-2 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

0.25-0.5 µg/mL
-

Immunocytochemistry (ICC/IF)

1-2 µg/mL
-

Flow Cytometry (Flow)

1-2 µg/10^6 cells
-
Product Specifications

Species Reactivity

Bovine, Human, Mouse, Non-human primate, Pig, Rat

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

HM57

Immunogen

A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Wet ice

Target Information

CD79a is a disulphide-linked heterodimer that includes B29 (CD79b) polypeptide. CD79a is a B lymphocyte antigen receptor with an antigen-specific surface component Ig (immunoglobulin) that associates with Ig-alpha and Ig-beta, necessary elements for the expression and function of the B-cell antigen receptor. CD79a first appears at pre-B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Diseases associated with CD79a include Agammaglobulinemia 3 and Agammaglobulinemia, Non-Bruton type.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79-alpha protein; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule, immunoglobulin-associated alpha; Ig alpha; Ig-alpha; immunoglobulin-associated alpha; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; Surface IgM-associated protein

View more View less

Gene Aliases: BOS_17604; CD79A; Ig-alpha; IGA; Igalpha; Ly-54; Ly54; MB-1; MB1

View more View less

UniProt ID: (Bovine) P40293, (Human) P11912, (Mouse) P11911

View more View less

Entrez Gene ID: (Bovine) 281674, (Human) 973, (Pig) 100190992, (Mouse) 12518, (Rat) 100913063

View more View less

Function(s)
transmembrane signaling receptor activity protein binding
Process(es)
cell surface receptor signaling pathway B cell differentiation B cell proliferation B cell activation B cell receptor signaling pathway adaptive immune response immune system process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-b648c57dc-dsdl2:80/100.66.129.203:80.
git-commit: 561b23eae95443db19074e56c849bd303926a883
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0