Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD72 Antibodies

Proteintech

CD72 Monoclonal Antibody (2C1C6)

View all (23) CD72 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD72 Monoclonal Antibody (2C1C6)

  • Antibody Testing Data (5)
CD72 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
CD72 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD72 Antibody (67424-1-IG) in IHC (P)

Immunohistochemical analysis of paraffin-embedded human tonsilitis tissue slide using 67424-1-Ig (CD72 antibody) at dilution of 1:1000 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD72 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CD72 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CD72 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CD72 Antibody in Western Blot (WB)
CD72 Antibody in Flow Cytometry (Flow)

Product Details

67424-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:10,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:1,000
-

Flow Cytometry (Flow)

0.20 µg
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

2C1C6

Immunogen

CD72 Fusion Protein Ag14405

Conjugate

Unconjugated Unconjugated Unconjugated
  • CoraLite 594
  • Request custom conjugation

Form

Liquid

Concentration

1000 µg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Aliquoting is unnecessary for -20°C storage.

Target Information

CD72 is a transmembrane glycoprotein expressed as a homodimer especially in B cells, but also in other antigen presenting cells such as dendritic cells and macrophages. Through one of its immunoreceptor tyrosine-based inhibitory motives (ITIMs), CD72 interacts with tyrosine phosphatase SHP-1, thereby suppressing B cell responsiveness. Binding of CD72 with its ligand CD100 (Sema4D) prevents BCR association and phosphorylation of CD72 and results in dissociation of SHP-1 from CD72, thus enables B cell activation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: B-cell differentiation antigen CD72; CD72; CD72 antigen; Lyb-2; RP11-331F9.9

View more View less

Gene Aliases: CD72; CD72b; LYB2

View more View less

UniProt ID: (Human) P21854

View more View less

Entrez Gene ID: (Human) 971

View more View less

Function(s)
transmembrane signaling receptor activity receptor binding protein binding carbohydrate binding
Process(es)
cell adhesion signal transduction
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0