Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD6 Antibodies

Invitrogen

CD6 Recombinant Rabbit Monoclonal Antibody (4H14L18)

Advanced Verification
View all (44) CD6 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD6 Recombinant Rabbit Monoclonal Antibody (4H14L18)

  • Antibody Testing Data (2)
  • Advanced Verification (1)
CD6 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
CD6 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD6 Antibody (702907) in WB

Western blot analysis was performed on membrane enriched extracts (30 µg lysate) of K-562 (Lane 1), Raji (Lane 2), HT-29 (Lane 3) and MCF-7 (Lane 4). The blot was probed with Anti-CD6 Recombinant Rabbit Monoclonal Antibody (Product # 702907, 1:5,000 dilution) and detected by chemiluminescence using Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, HRP conjugate (Product # A27036, 0.25 µg/mL, 1:4,000 dilution). A 72 Da band corresponding to CD6 was observed across the cell lines tested. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD6 Antibody in Western Blot (WB)
CD6 Antibody in Western Blot (WB)
CD6 Antibody
CD6 Recombinant Rabbit Monoclonal Antibody (4H14L18)

Product Details

702907

Applications
Tested Dilution
Publications

Western Blot (WB)

1:5,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

Expi293

Class

Recombinant Monoclonal

Type

Antibody

Clone

4H14L18

Immunogen

Peptides corresponding to human CD6 [1)aa220-aa237 2)aa641-aa658]
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

0.09% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Wet ice

RRID

AB_2762403

Product Specific Information

This antibody is predicted to react with Monkey, Dog, Rabbit.

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Target Information

CD6, also known as T12, is a member of the scavenger receptor superfamily found on T and B cell subsets, thymocytes, and acute lymphocytic leukemia cells (ALL). CD6 interacts with its ligand CD166/ALCAM (activated leukocyte cell adhesion molecule) and serves as a coreceptor for T cell activation and stabilizer of the immunological synapse. CD6-ALCAM mediated cell adhesion is also important for T cell proliferation. CD6 may exert some its functions via association with CD5, probably by fine-tuning CD5 signaling. Ligation of CD6 has antiapoptotic role in chronic lymphocytic leukemia B cells.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD6; CD6 antigen; FLJ44171; T-cell differentiation antigen CD6; T12; TP120

View more View less

Gene Aliases: CD6; TP120

View more View less

UniProt ID: (Human) P30203

View more View less

Entrez Gene ID: (Human) 923

View more View less

Function(s)
lipopolysaccharide binding scavenger receptor activity protein binding lipoteichoic acid binding
Process(es)
immunological synapse formation adaptive immune response acute inflammatory response to antigenic stimulus receptor-mediated endocytosis heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules lipopolysaccharide-mediated signaling pathway response to lipopolysaccharide positive regulation of T cell proliferation innate immune response positive regulation of cytokine production involved in inflammatory response
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0