Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD5 Antibodies

Proteintech

CD5 Polyclonal Antibody

3 References
View all (295) CD5 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD5 Polyclonal Antibody

  • Antibody Testing Data (5)
CD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
CD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD5 Antibody (17227-1-AP) in IHC (P)

Immunohistochemistry of paraffin-embedded human tonsillitis tissue slide using 17227-1-AP (CD5 antibody) at dilution of 1:800 (under 40x lens) heat mediated antigen retrieved with Tris-EDTA buffer (pH 9). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CD5 Antibody in Western Blot (WB)
CD5 Antibody in Western Blot (WB)
CD5 Antibody in Immunoprecipitation (IP)
CD5 Polyclonal Antibody

Product Details

17227-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
View 1 publication 1 publication

Immunohistochemistry (IHC)

-
View 2 publications 2 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:400-1:1,600
-

Immunoprecipitation (IP)

0.5-4.0 µg
-
Product Specifications

Species Reactivity

Human, Mouse

Published species

Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

CD5 Fusion Protein Ag11065 (30-378 aa encoded by BC027901)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.4 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: DPDFQARLT RSNSKCQGQL EVYLKDGWHM VCSQSWGRSS KQWEDPSQAS KVCQRLNCGV PLSLGPFLVT YTPQSSIICY GQLGSFSNCS HSRNDMCHSL GLTCLEPQKT TPPTTRPPPT TTPEPTAPPR LQLVAQSGGQ HCAGVVEFYS GSLGGTISYE AQDKTQDLEN FLCNNLQCGS FLKHLPETEA GRAQDPGEPR EHQPLPIQWK IQNSSCTSLE HCFRKIKPQK SGRVLALLCS GFQPKVQSRL VGGSSICEGT VEVRQGAQWA ALCDSSSARS SLRWEEVCRE QQCGSVNSYR VLDAGDPTSR GLFCPHQKLS QCHELWERNS YCKKVFVTCQ DPNPAGLAAG (30-378 aa encoded by BC027901)

Target Information

CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter's Syndrome.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: CD antigen CD5; CD5; CD5 antigen (p56 62); CD5 antigen (p56-62); CD5 antigen p56-62; fCD5; Ly-1; Ly12; LyA; Lymphocyte antigen 1; Lymphocyte antigen T1/Leu-1; Pan T cell; T-cell surface glycoprotein CD5

View more View less

Gene Aliases: CD5; LEU1; Ly-1; Ly-12; Ly-A; Lyt-1; T1

View more View less

UniProt ID: (Human) P06127, (Mouse) P13379

View more View less

Entrez Gene ID: (Human) 921, (Mouse) 12507

View more View less

Function(s)
receptor activity transmembrane signaling receptor activity scavenger receptor activity protein binding glycoprotein binding
Process(es)
receptor-mediated endocytosis cell recognition cell proliferation T cell costimulation apoptotic signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0