Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD47 Antibodies

Invitrogen

CD47 Monoclonal Antibody (B6H12), APC-eFluor™ 780, eBioscience™

9 Published Figures
9 References
View all (156) CD47 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD47 Monoclonal Antibody (B6H12), APC-eFluor™ 780, eBioscience™

  • Antibody Testing Data (1)
  • Published Figures (9)
CD47 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD47 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 10

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD47 Antibody (47-0479-42) in Flow

Normal human peripheral blood cells were stained with Mouse IgG1 kappa Isotype Control, APC-eFluor 780 (Product # 47-4714-82) (blue histogram) or CD47 Monoclonal Antibody, APC-eFluor 780 (purple histogram). Viable cells in the lymphocyte gate were used for analysis, as determined by Fixable Viability Dye eFluor 450 (Product # 65-0863-18). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD47 Antibody in Flow Cytometry (Flow)
CD47 Antibody in Flow Cytometry (Flow)
CD47 Antibody in Flow Cytometry (Flow)
CD47 Antibody in Flow Cytometry (Flow)
CD47 Antibody in Flow Cytometry (Flow)
CD47 Antibody in Flow Cytometry (Flow)
CD47 Antibody in Flow Cytometry (Flow)
CD47 Antibody in Flow Cytometry (Flow)
CD47 Antibody in Inhibition Assays (IA)
CD47 Antibody in Inhibition Assays (IA)

Product Details

47-0479-42

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

-
View 1 publication 1 publication

Flow Cytometry (Flow)

5 µL (0.5 µg)/test
View 6 publications 6 publications

Inhibition Assays (IA)

-
View 2 publications 2 publications
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Recommended Isotype Control

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), APC-eFluor™ 780, eBioscience™

Class

Monoclonal

Type

Antibody

Clone

B6H12

Conjugate

APC-eFluor™ 780 APC-eFluor™ 780 APC-eFluor™ 780
  • Unconjugated
  • Alexa Fluor 488 
  • Alexa Fluor 700 
  • APC 
  • eFluor 450
  • FITC
  • Functional Grade
  • NovaFluor Blue 660-120S
  • NovaFluor Red 700
  • NovaFluor Red 725
  • NovaFluor Red 755
  • NovaFluor Yellow 610
  • NovaFluor Yellow 660
  • PE-Cyanine5
  • PE-Cyanine7
  • PerCP-eFluor 710
  • Request custom conjugation

Excitation/Emission Max

756/785 nm View spectra spectra

Form

Liquid

Concentration

5 µL/Test

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2, with 0.2% BSA

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2811816

Product Specific Information

Description: The monoclonal antibody B6H12 reacts to CD47 also known as integrin-associated protein (IAP), and neurophilin. CD47 is a glycosylated five transmembrane protein with a small alternatively spliced cytoplasmic domain. CD47 is involved in adhesion through interactions with SIRP (signal regulator protein) and is non-covalently associated with beta3 integrins CD51/CD61 and CD41/CD61. Furthermore this interaction can mediate bi-directional signaling to modify neural synaptic activity and regulate the phagocytic activities of macrophages. CD47 is the receptor for thrombospondin. T cell expression of CD47 can mediate activation or apoptosis (in the presence of high levels of thrombospondin). Recently stimulation of CD47 by monoclonal antibody has been shown to induce CD4+CD25- suppressive activity also increasing expression of Foxp3. Expression is found in the majority of hematopoietic cells including T and B cells, monocytes, platelets and erythrocytes (as part of the Rh complex). Expression is also found in non-hematopoietic cells.

This antibody has been reported to have neutralizing activity.

Applications Reported: This B6H12 antibody has been reported for use in flow cytometric analysis.

Applications Tested: This B6H12 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633-647 nm). Please make sure that your instrument is capable of detecting this fluorochrome.

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser.

Target Information

CD47, known as integrin-associated protein (IAP), is a glycosylated transmembrane glycoprotein with five domains, expressed widely across hematopoietic cells like T and B cells, monocytes, platelets, and erythrocytes, as well as non-hematopoietic cells. It interacts with integrins such as CD51/CD61 and CD41/CD61, and serves as a receptor for thrombospondin, mediating bi-directional signaling that affects neural synaptic activity and macrophage phagocytosis. CD47 acts as a ligand for CD172a (SIRP alpha), an inhibitory receptor on macrophages, preventing phagocytosis of CD47-positive cells. This interaction influences cell migration, B cell adhesion, T cell activation, and neuronal development, particularly in synapse-rich brain and retina regions. It also modulates chondrocyte responses to mechanical signals. T cell expression of CD47 can lead to activation or apoptosis in the presence of thrombospondin. Monoclonal antibody stimulation of CD47 has been shown to induce CD4+CD25- suppressive activity and increase Foxp3 expression. CD47's role in membrane transport, signal transduction, and its broad tissue distribution highlight its significance in various physiological processes.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

Bioinformatics

Protein Aliases: antigen identified by monoclonal antibody 1D8; Antigenic surface determinant protein OA3; CD47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 glycoprotein; IAP; integrin associated protein; Integrin-associated protein; integrin-associated signal transducer; Leukocyte surface antigen CD47; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47

View more View less

Gene Aliases: CD47; IAP; MER6; OA3

View more View less

UniProt ID: (Human) Q08722

View more View less

Entrez Gene ID: (Human) 961

View more View less

Function(s)
protein binding thrombospondin receptor activity
Process(es)
cell adhesion integrin-mediated signaling pathway opsonization positive regulation of cell proliferation response to bacterium positive regulation of cell-cell adhesion extracellular matrix organization positive regulation of inflammatory response positive regulation of phagocytosis positive regulation of T cell activation leukocyte migration
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0