Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • KIM-1 Antibodies

Invitrogen

CD365 (Tim-1) Recombinant Rabbit Monoclonal Antibody (P365D), Alexa Fluor™ 647, eBioscience™

Advanced Verification
View all (41) KIM-1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD365 (Tim-1) Recombinant Rabbit Monoclonal Antibody (P365D), Alexa Fluor™ 647, eBioscience™

  • Antibody Testing Data (3)
  • Advanced Verification (1)
CD365 (Tim-1) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD365 (Tim-1) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD365 (Tim-1) Antibody (51-3658-42) in Flow

Antibody specificity was demonstrated by siRNA mediated knockdown of CD365. 769P cells were transfected with either non-targeting control siRNA or siRNA against CD365. As expected, loss of signal was observed with CD365 clone P365D in cells transfected with the CD365 specific siRNA (pink histogram) but not the cells transfected with control siRNA (purple histogram). SI denotes the staining index calculated against corresponding isotype controls. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD365 (Tim-1) Antibody in Flow Cytometry (Flow)
CD365 (Tim-1) Antibody in Flow Cytometry (Flow)
CD365 (Tim-1) Antibody in Flow Cytometry (Flow)
CD365 (Tim-1) Antibody
CD365 (Tim-1) Recombinant Rabbit Monoclonal Antibody (P365D), Alexa Fluor™ 647, eBioscience™

Product Details

51-3658-42

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

5 µL (0.06 µg)/test
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

Expi293

Class

Recombinant Monoclonal

Type

Antibody

Clone

P365D

Immunogen

Protein corresponding to Human CD365 (aa22-aa129)
View immunogen

Conjugate

Alexa Fluor™ 647 Alexa Fluor™ 647 Alexa Fluor™ 647
  • Unconjugated
  • Alexa Fluor 488 
  • Request custom conjugation

Excitation/Emission Max

650/671 nm View spectra spectra

Form

Liquid

Concentration

5 µL/Test

Purification

Protein A

Storage buffer

proprietary buffer/PBS, pH 7.2

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Wet ice

RRID

AB_2848405

Product Specific Information

Description: This P365D recombinant rabbit monoclonal antibody reacts with human CD365, which is also known as Tim-1, Kim-1, or Havcr1.

Applications Tested: This P365D antibody has been pre-diluted and tested by flow cytometric analysis of 769P cells. This may be used at 5 µL (0.06 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser

Filtration: 0.2 µm post-manufacturing filtered.

Target Information

CD365 also known as T-cell immunoglobulin and mucin domain 1 (Tim-1) is a type 1 surface glycoprotein that consists of extracellular N-terminal immunoglobulin V-like domain and a mucin domain, and a short cytoplasmic tail. The endogenous ligand for CD365 is Tim-4. In addition, CD365 is known to be a cellular receptor or co-receptor for a number of viruses including hepatitis A and C viruses, Ebola virus, Marburg virus, Dengue virus, Zika virus, West Nile virus, and Japanese encephalitis virus. The direct binding of the viruses by CD365 occurs via phosphatildyl-serine exposed on viral envelopes. Polymorphisms within human CD365 gene (HAVCR1) are associated with susceptibility to infection and the disease severity. CD365 has also been implicated in the T cell co-stimulation. CD365 is primarily expressed in epithelial cells. It is also highly expressed in A549, 769-P and Caco-2 and cell lines. CD365 exists also as a soluble protein generated by metalloproteinase mediated proteolytic cleavage of the surface protein.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD365; HAVcr-1; Hepatitis A virus cellular receptor 1; Kidney injury molecule 1; KIM-1; T cell immunoglobin domain and mucin domain protein 1; T-cell immunoglobulin and mucin domain-containing protein 1; T-cell immunoglobulin mucin family member 1; T-cell immunoglobulin mucin receptor 1; T-cell membrane protein 1; TIM; TIMD-1

View more View less

Gene Aliases: CD365; HAVCR; HAVCR-1; HAVCR1; KIM-1; KIM1; TIM; TIM-1; TIM1; TIMD-1; TIMD1

View more View less

UniProt ID: (Human) Q96D42

View more View less

Entrez Gene ID: (Human) 26762

View more View less

Function(s)
virus receptor activity
Process(es)
viral entry into host cell
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0