Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • DPP4 Antibodies

NeoBiotechnologies

CD26 (DPP IV/ADA-Binding Protein) Monoclonal Antibody (DPP4/7415)

View all (68) DPP4 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD26 (DPP IV/ADA-Binding Protein) Monoclonal Antibody (DPP4/7415)

  • Antibody Testing Data (3)
CD26 (DPP IV/ADA-Binding Protein) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
CD26 (DPP IV/ADA-Binding Protein) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD26 (DPP IV/ADA-Binding Protein) Antibody (1803-MSM5-P1) in IHC (P)

Formalin-fixed, paraffin-embedded human prostate cancer stained with CD26 Mouse Monoclonal Antibody (DPP4/7415). HIER: Tris/EDTA, pH9.0, 45 min. 2°C: HRP-polymer, 30 min. DAB, 5 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD26 (DPP IV/ADA-Binding Protein) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CD26 (DPP IV/ADA-Binding Protein) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
CD26 (DPP IV/ADA-Binding Protein) Antibody in Western Blot (WB)
CD26 (DPP IV/ADA-Binding Protein) Monoclonal Antibody (DPP4/7415)

Product Details

1803-MSM5-P1

Applications
Tested Dilution
Publications

Western Blot (WB)

2-4 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

1-2 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2, kappa

Class

Monoclonal

Type

Antibody

Clone

DPP4/7415

Immunogen

Recombinant fragment (around aa1-200) of human DPP4 protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Positive Control:Human lymph nodes or tonsil.

Cellular Location: Cell surface

Target Information

CD26 (dipeptidyl peptidase IV, DPP IV), adenosine deaminase (ADA) binding protein) is a homodimeric atypical serine protease belonging to the prolyl oligopeptidase family. CD26 is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. CD26 identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. Further, CD26 is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. Alterations in CD26 peptidase activity are characteristic of malignant transformation, and the enzymatic activity increases dramatically with tumor grade and severity. CD26 is expressed in various blood cell types, but also in cells that are histogenetically related to activated fibroblasts. Alterations in CD26 density have been reported on circulating monocytes and CD4+ T cells during rheumatoid arthritis and systemic lupus erythematosus.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: ADABP; ADCP-2; Adenosine deaminase complexing protein 2; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase IV; dipeptidyl-peptidase 4; dipeptidylpeptidase 4; dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP-IV; T-cell activation antigen CD26; TP103

View more View less

Gene Aliases: ADABP; ADCP2; CD26; DPP4; DPPIV; TP103

View more View less

UniProt ID: (Human) P27487

View more View less

Entrez Gene ID: (Human) 1803

View more View less

Function(s)
virus receptor activity protease binding serine-type endopeptidase activity receptor binding protein binding serine-type peptidase activity dipeptidyl-peptidase activity identical protein binding protein homodimerization activity
Process(es)
behavioral fear response response to hypoxia proteolysis positive regulation of cell proliferation negative regulation of extracellular matrix disassembly T cell costimulation regulation of cell-cell adhesion mediated by integrin locomotory exploration behavior psychomotor behavior T cell activation endothelial cell migration viral entry into host cell
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-585975bbdc-x5cgg:80/100.66.128.25:80.
git-commit: a17be567db00e29ba041ba55be3678c43488a11a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0