Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD1d Antibodies

Proteintech

CD1d Monoclonal Antibody (1B1), CoraLite® Plus 647

View all (45) CD1d antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD1d Monoclonal Antibody (1B1), CoraLite® Plus 647

  • Antibody Testing Data (1)
CD1d Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD1d Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD1d Antibody (CL647-65242) in Flow

1X10^6 mouse splenocytes were surface stained with FITC Anti-Mouse CD45R (B220) (FITC-65139, Clone: RA3-6B2) and 1 µg CoraLite® Plus 647 Anti-Mouse CD1d (CL647-65242, Clone: 1B1) or CoraLite® Plus 647 conjugated rat IgG2b isotype control. Cells were not fixed. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD1d Antibody in Flow Cytometry (Flow)
CD1d Monoclonal Antibody (1B1), CoraLite® Plus 647

Product Details

CL647-65242

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

Assay-dependent
-
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Rat / IgG2b, kappa

Class

Monoclonal

Type

Antibody

Clone

1B1

Immunogen

Mouse CD1d cDNA-transfected RMA-S mouse T lymphoma
View immunogen

Conjugate

CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647
  • Unconjugated
  • CoraLite 594
  • CoraLite Plus 488
  • CoraLitePlus 405
  • FITC Plus
  • Request custom conjugation

Excitation/Emission Max

654/674 nm

Form

Liquid

Purification

Affinity chromatography

Storage buffer

PBS

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark

Shipping conditions

Wet ice

Target Information

CD1d is a heavy chain associated with Beta 2 microglobulin on cortical thymocytes. Beta 2 microglobulin independent expression of CD1d has also been demonstrated on human intestinal epithelial cells. CD1d is the sole group 2 member of the CD1 family of major histocompatibility (MHC) like glycoproteins. The CD1d gene encodes a divergent member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by the CD1d gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail. Diseases associated with CD1D include Mycobacterium Malmoense and Autoimmune Disease.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Antigen-presenting glycoprotein CD1d1; CD1d.1; T-cell surface glycoprotein CD1d1

View more View less

Gene Aliases: AI747460; CD1.1; Cd1a; Cd1d; Cd1d1; Ly-38

View more View less

UniProt ID: (Mouse) P11609

View more View less

Entrez Gene ID: (Mouse) 12479

View more View less

Function(s)
protein binding beta-2-microglobulin binding lipid antigen binding endogenous lipid antigen binding exogenous lipid antigen binding histone binding T cell receptor binding cell adhesion molecule binding lipopeptide binding
Process(es)
positive regulation of T cell mediated cytotoxicity immune system process immune response antigen processing and presentation positive regulation of T cell proliferation positive regulation of macrophage activation positive thymic T cell selection positive regulation of interferon-gamma biosynthetic process positive regulation of interleukin-2 biosynthetic process innate immune response positive regulation of interleukin-4 biosynthetic process antigen processing and presentation, endogenous lipid antigen via MHC class Ib antigen processing and presentation, exogenous lipid antigen via MHC class Ib regulation of immune response positive regulation of NK T cell activation positive regulation of NK T cell differentiation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0