Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CXCR5 Antibodies

Invitrogen

CD185 (CXCR5) Monoclonal Antibody (MU5UBEE), PE, eBioscience™

15 References
View all (62) CXCR5 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD185 (CXCR5) Monoclonal Antibody (MU5UBEE), PE, eBioscience™

  • Antibody Testing Data (1)
CD185 (CXCR5) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD185 (CXCR5) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD185 (CXCR5) Antibody (12-9185-42) in Flow

Staining of normal human peripheral blood cells with Anti-Human CD19 APC (Product # 17-0199-42) and Mouse IgG2b K Isotype Control PE (Product # 12-4732-81) (left) or Anti-Human CD185 (CXCR5) PE (right). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD185 (CXCR5) Antibody in Flow Cytometry (Flow)
CD185 (CXCR5) Monoclonal Antibody (MU5UBEE), PE, eBioscience™

Product Details

12-9185-42

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

5 µL (0.125 µg)/test
View 15 publications 15 publications
Product Specifications

Species Reactivity

Human, Non-human primate, Rhesus monkey

Published species

Human, Non-human primate, Rhesus monkey

Host/Isotype

Mouse / IgG2b, kappa

Recommended Isotype Control

Mouse IgG2b kappa Isotype Control (eBMG2b), PE, eBioscience™

Class

Monoclonal

Type

Antibody

Clone

MU5UBEE

Immunogen

N-terminal extracellular domain
View immunogen

Conjugate

PE PE PE
  • Unconjugated
  • Alexa Fluor 532 
  • Alexa Fluor 700 
  • APC 
  • Biotin 
  • Brilliant UV 395
  • Brilliant UV 615
  • Brilliant UV 737
  • Brilliant UV 805
  • Brilliant Violet 421
  • Brilliant Violet 480
  • Brilliant Violet 605
  • Brilliant Violet 786
  • eFluor 450
  • FITC
  • NovaFluor Blue 610-70S
  • NovaFluor Blue 660-120S
  • NovaFluor Blue 725
  • NovaFluor Blue 760
  • NovaFluor Blue 800
  • NovaFluor Red 685
  • NovaFluor Red 710
  • NovaFluor UV 765
  • NovaFluor Violet 690
  • NovaFluor Violet 800
  • NovaFluor Yellow 610
  • NovaFluor Yellow 690
  • NovaFluor Yellow 730
  • NovaFluor Yellow 810
  • PE-Cyanine5
  • PE-Cyanine7
  • PE-eFluor 610
  • PerCP-eFluor 710
  • Super Bright 436
  • Super Bright 600
  • Super Bright 645
  • Super Bright 702
  • Request custom conjugation

Excitation/Emission Max

565/576 nm View spectra spectra

Form

Liquid

Concentration

5 µL/Test

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2, with BSA

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_11219877

Product Specific Information

Description: The MU5UBEE monoclonal antibody reacts with human and non-human primate CD185. CD185, which is also known as C-X-C chemokine receptor 5 (CXCR5) and Burkitt lymphoma receptor 1 (BLR1), is a seven transmembrane G protein-coupled receptor originally identified in Burkitt's lymphoma. In peripheral blood, CD185 is expressed on B cells, CD4+ T cells (but not Th1 or Th2 cells), as well as on a subpopulation of memory (CD45RO+) T cells. Circulating CD185+ T cells are in a resting state and migrate to the lymph nodes due to expression of CCR7 and CD62L. In tonsil, CD185 is expressed on nearly all CD4+ cells together with CD45RO and activation markers such as CD69 and ICOS. Tonsillar CD185+ cells have been shown to induce antibody production when co-cultured with B cells, thus supporting their role in providing help to B cells. Furthermore, this chemokine receptor plays a critical role in lymphocyte trafficking, in particular T cell migration into the B cell follicles of germinal centers in response to CXCL13, making CD185 an established marker of follicular helper T cells.

The MU5UBEE monoclonal antibody also crossreacts with Rhesus macaque.

Applications Reported: This MU5UBEE antibody has been reported for use in flow cytometric analysis.

Applications Tested: This MU5UBEE antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

Filtration: 0.2 µm post-manufacturing filtered.

Target Information

CD185, also known as CXCR5 or Burkitt lymphoma receptor 1 (BLR1), is a seven-transmembrane G protein-coupled receptor in the CXC chemokine receptor family. It plays a crucial role in lymphocyte trafficking, particularly in T cell migration into B cell follicles of germinal centers in response to CXCL13, making it a key marker of follicular helper T cells. In peripheral blood, CXCR5 is expressed on B cells, CD4+ T cells (excluding Th1 and Th2 cells), and a subset of memory (CD45RO+) T cells. These CD185+ T cells are in a resting state and migrate to lymph nodes due to CCR7 and CD62L expression. In tonsils, CD185 is found on nearly all CD4+ cells, along with activation markers like CD69 and ICOS, and can induce antibody production when co-cultured with B cells. CXCR5 is expressed in lymphatic tissues such as lymph node follicles and the spleen, and is also found in human blood and salivary glands, as well as various animal tissues. It is essential for B cell migration and is part of the larger family of G protein-coupled receptors (GPCRs), which have seven membrane-spanning helices and are involved in numerous signaling pathways. GPCRs, including CXCR5, are regulated by various ligands that affect their conformation and signaling through G-proteins and downstream effectors. The regulation of GPCRs, including their deactivation, has significant implications for cellular functions and is a major focus of pharmacological research, leading to the development of drugs like Zyprexa and Claritin for mental health and allergy treatments.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

Bioinformatics

Protein Aliases: Blr-1; Burkitt lymphoma receptor 1; Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif) receptor 5); Burkitt lymphoma receptor 1, GTP-binding protein; C Cmotif chemokine; C X C motif chemokine; C-X-C chemokine receptor type 5; CC motif chemokine; CCmotif chemokine; CD185; CD185 antigen; chemokine (C-X-C motif) receptor 5; CXC; CXC motif chemokine; CXC-R5; CXCR-5; Fusin; GPR9; HUMSTR; LESTR; MDR-15; Monocyte-derived receptor 15

View more View less

Gene Aliases: BLR1; CD185; CXCR5; EGK_06966; MDR15

View more View less

UniProt ID: (Human) P32302

View more View less

Entrez Gene ID: (Rhesus monkey) 701792, (Human) 643

View more View less

Function(s)
G-protein coupled receptor activity protein binding C-X-C chemokine receptor activity
Process(es)
movement of cell or subcellular component chemotaxis immune response G-protein coupled receptor signaling pathway positive regulation of cytokinesis B cell activation lymph node development chemokine-mediated signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0