Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD14 Antibodies

Invitrogen

CD14 Monoclonal Antibody (61D3), Brilliant Violet™ 605, eBioscience™

View all (206) CD14 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD14 Monoclonal Antibody (61D3), Brilliant Violet™ 605, eBioscience™

  • Antibody Testing Data (1)
CD14 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD14 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD14 Antibody (406-0149-42) in Flow

Normal human lysed whole blood cells were stained with Mouse IgG1 kappa Isotype Control, Brilliant Violet 605 (Product # 406-4714-81) (blue histogram) or CD14 Monoclonal Antibody, Brilliant Violet 605 (purple histogram). Viable cells in the monocyte gate were used for analysis, as determined by 7-AAD (Product # 00-6993-50). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD14 Antibody in Flow Cytometry (Flow)

Product Details

406-0149-42

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

5 µL (0.5 µg)/test
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Recommended Isotype Control

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), Brilliant Violet™ 605, eBioscience™

Class

Monoclonal

Type

Antibody

Clone

61D3

Conjugate

Brilliant Violet™ 605 Brilliant Violet™ 605 Brilliant Violet™ 605
  • Unconjugated
  • Alexa Fluor 488 
  • Alexa Fluor 532 
  • Alexa Fluor 660 
  • Alexa Fluor 700 
  • APC 
  • APC-eFluor 780
  • Biotin 
  • Brilliant UV 395
  • Brilliant UV 496
  • Brilliant UV 615
  • Brilliant UV 661
  • Brilliant UV 737
  • Brilliant UV 805
  • Brilliant Violet 421
  • Brilliant Violet 480
  • Brilliant Violet 650
  • Brilliant Violet 711
  • Brilliant Violet 786
  • eFluor 450
  • eFluor 506
  • FITC
  • Functional Grade
  • NovaFluor Blue 510
  • NovaFluor Blue 555
  • NovaFluor Blue 585
  • NovaFluor Blue 610-30S
  • NovaFluor Blue 660-40S
  • NovaFluor Blue 800
  • NovaFluor Yellow 570
  • NovaFluor Yellow 660
  • PE 
  • PE-Cyanine5
  • PE-Cyanine7
  • PE-eFluor 610
  • PerCP-Cyanine5.5
  • PerCP-eFluor 710
  • Super Bright 436
  • Super Bright 600
  • Super Bright 645
  • Super Bright 702
  • Super Bright 780
  • Request custom conjugation

Excitation/Emission Max

406/603 nm View spectra spectra

Form

Liquid

Concentration

5 µL/Test

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2, with BSA

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3093236

Product Specific Information

Description
The 61D3 monoclonal antibody reacts with human CD14, a 53-55 kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.

Applications Tested
This 61D3 antibody has been pre-diluted and tested by flow cytometric analysis of normal human lysed whole blood cells. This may be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

Blocking Buffers
When using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) or Brilliant Stain Buffer (Product # 00-4409-75) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer or Brilliant Stain Buffer for more information.

Light sensitivity
This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

Fixation
• Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
• Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
• Our internal testing suggests that Brilliant Violet™ 605 (BV605) is not compatible with methanol-based fixation.

Excitation: 407 nm; Emission: 605 nm; Laser: Violet Laser.

BRILLIANT ULTRA VIOLET™ is a trademark or registered trademark of Becton, Dickinson and Company or its affiliates, and is used under license. Powered by Sirigen™.

Target Information

CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD 14; CD14; cd14 monocyte; LPSR antibody; Monocyte differentiation antigen CD14; My23 antigen; Myeloid cell-specific leucine-rich glycoprotein; sCD14; soluble CD14

View more View less

Gene Aliases: CD14

View more View less

UniProt ID: (Human) P08571

View more View less

Entrez Gene ID: (Human) 929

View more View less

Function(s)
lipopolysaccharide binding opsonin receptor activity protein binding peptidoglycan receptor activity lipoteichoic acid binding
Process(es)
MyD88-dependent toll-like receptor signaling pathway MyD88-independent toll-like receptor signaling pathway receptor-mediated endocytosis phagocytosis apoptotic process inflammatory response cell surface receptor signaling pathway I-kappaB kinase/NF-kappaB signaling response to heat lipopolysaccharide-mediated signaling pathway response to magnesium ion positive regulation of type I interferon production positive regulation of interferon-gamma production positive regulation of tumor necrosis factor production negative regulation of MyD88-independent toll-like receptor signaling pathway toll-like receptor 4 signaling pathway response to tumor necrosis factor TRIF-dependent toll-like receptor signaling pathway toll-like receptor TLR1:TLR2 signaling pathway toll-like receptor TLR6:TLR2 signaling pathway innate immune response response to ethanol positive regulation of endocytosis positive regulation of cytokine secretion response to electrical stimulus necroptotic process cellular response to lipopolysaccharide cellular response to lipoteichoic acid cellular response to diacyl bacterial lipopeptide cellular response to triacyl bacterial lipopeptide apoptotic signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0