Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD11b Antibodies

Invitrogen

CD11b Monoclonal Antibody (ICRF44), PE-eFluor™ 610, eBioscience™

14 Published Figures
18 References
View all (219) CD11b antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD11b Monoclonal Antibody (ICRF44), PE-eFluor™ 610, eBioscience™

  • Antibody Testing Data (1)
  • Published Figures (14)
CD11b Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD11b Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 15

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD11b Antibody (61-0118-42) in Flow

Normal human peripheral blood cells were stained with Mouse IgG1 kappa Isotype Control, PE-eFluor 610 (Product # 61-4714-82) (blue histogram) or CD11b Monoclonal Antibody, PE-eFluor 610 (purple histogram). Cells in the monocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Immunohistochemistry (IHC)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)
CD11b Antibody in Flow Cytometry (Flow)

Product Details

61-0118-42

Applications
Tested Dilution
Publications

Immunohistochemistry (IHC)

-
View 1 publication 1 publication

Flow Cytometry (Flow)

5 µL (0.5 µg)/test
View 17 publications 17 publications
Product Specifications

Species Reactivity

Baboon, Chimpanzee, Cynomolgus monkey, Human, Rhesus monkey

Host/Isotype

Mouse / IgG1, kappa

Recommended Isotype Control

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PE-eFluor™ 610, eBioscience™

Class

Monoclonal

Type

Antibody

Clone

ICRF44

Conjugate

PE-eFluor™ 610 PE-eFluor™ 610 PE-eFluor™ 610
  • Unconjugated
  • Alexa Fluor 700 
  • APC 
  • APC-eFluor 780
  • Biotin 
  • Brilliant UV 496
  • Brilliant UV 563
  • Brilliant UV 615
  • Brilliant UV 661
  • Brilliant UV 737
  • Brilliant UV 805
  • Brilliant Violet 421
  • Brilliant Violet 605
  • Brilliant Violet 650
  • Brilliant Violet 711
  • eFluor 506
  • FITC
  • NovaFluor Blue 610-70S
  • NovaFluor Blue 660-120S
  • NovaFluor Blue 800
  • NovaFluor Red 710
  • NovaFluor Red 725
  • NovaFluor Red 755
  • NovaFluor Violet 690
  • NovaFluor Violet 800
  • NovaFluor Yellow 570
  • NovaFluor Yellow 590
  • NovaFluor Yellow 660
  • NovaFluor Yellow 755
  • NovaFluor Yellow 810
  • PE 
  • PE-Cyanine5
  • PE-Cyanine7
  • PerCP-eFluor 710
  • Super Bright 436
  • Super Bright 600
  • Super Bright 645
  • Super Bright 702
  • Super Bright 780
  • Request custom conjugation

Excitation/Emission Max

565/606 nm View spectra spectra

Form

Liquid

Concentration

5 µL/Test

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2, with 0.2% BSA

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2802371

Product Specific Information

Description: The ICRF44 monoclonal antibody reacts with human CD11b, a 165 kDa adhesion molecule. CD11b associated with integrin beta2 (CD18) is expressed on the surface of monocytes, granulocytes, activated lymphocytes and a subset of NK cells. CD11b is a receptor for intercellular adhesion molecule family members CD54, CD102 and CD50 as well as for iC3b. These adhesions are crucial in cell-cell and cell-matrix interactions.

Applications Reported: This ICRF44 antibody has been reported for use in flow cytometric analysis.

Applications Tested: This ICRF44 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

PE-eFluor 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red). Please make sure that your instrument is capable of detecting this fluorochrome.

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

Target Information

CD11b (integrin alpha-M, ITGAM, integrin alpha-X, ITGAX) is a 165 kDa adhesion molecule that associates non-covalently with integrin beta-2 (CD18). The CD11b/CD18 heterodimeric complex is also known as integrin alpha-M beta-2, Mac-1, and CR3 (complement receptor 3). CD11b is expressed on the surface of monocytes/macrophages, granulocytes, activated lymphocytes, a subset of NK cells, a subset of dendritic cells, and microglia in the brain. CD11b/CD18 functions as the receptor for ICAM-1 (CD54), ICAM-2 (CD102), ICAM-4 (CD242), CD14, CD50, CD23, heparin, iC3b, fibrinogen, and Factor X -these adhesions are critical for cell-cell and cell-matrix interactions. CD11b is expressed on 8% of spleen cells, 44% of bone marrow cells, and less than 1% of thymocytes, and is commonly used as a microglial marker in nervous tissue. The expression of CD11b increases during monocyte maturation and expression levels vary on tissue macrophages. Further, peritoneal macrophages are reported to express higher levels of CD11b than splenic macrophages. Diseases associated with CD11b dysfunction include systemic lupus erythematosus 6 and ITGAM-related susceptibility to systemic lupus erythematosus.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

Bioinformatics

Protein Aliases: antigen CD11b (p170); CD11 antigen-like family member B; CD11b; Cell surface glycoprotein MAC-1 subunit alpha; complement component 3 receptor 3 subunit; CR-3 alpha chain; integrin alpha M; Integrin alpha-M; integrin, alpha M (complement component 3 receptor 3 subunit); Leukocyte adhesion receptor MO1; Mac-1 alpha (Mac1A); MAC-1 alpha subunit; macrophage antigen alpha polypeptide; MGC117044; Neutrophil adherence receptor; neutrophil adherence receptor alpha-M subunit; uncharacterized LOC101866035

View more View less

Gene Aliases: CD11B; CR3A; ITGAM; MAC-1; MAC1A; MO1A; SLEB6

View more View less

UniProt ID: (Human) P11215

View more View less

Entrez Gene ID: (Human) 3684, (Chimpanzee) 454069, (Cynomolgus monkey) 101866035

View more View less

Function(s)
glycoprotein binding protein binding metal ion binding
Process(es)
cell adhesion integrin-mediated signaling pathway ectodermal cell differentiation extracellular matrix organization toll-like receptor 4 signaling pathway leukocyte migration
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0