Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD11a Antibodies

Invitrogen

CD11a (LFA-1alpha) Monoclonal Antibody (BL1H8), FITC

View all (116) CD11a antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD11a (LFA-1alpha) Monoclonal Antibody (BL1H8), FITC

  • Antibody Testing Data (1)
CD11a (LFA-1alpha) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD11a (LFA-1alpha) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD11a (LFA-1alpha) Antibody (MA5-28271) in Flow

Flow cytometry of CD11a (LFA-1alpha) in red cell lysed porcine blood. Sample was incubated with CD11a (LFA-1alpha) monoclonal antibody (Product # MA5-28271) and gated on mononuclear cells. Figures are arranged as follows: A) FITC conjugated mouse anti porcine CD11a and purified mouse IgG1 isotype control detected with goat anti mouse IgG1 PE. B) FITC conjugated mouse anti porcine CD11a and purified mouse an... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD11a (LFA-1alpha) Antibody in Flow Cytometry (Flow)

Product Details

MA5-28271

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

Assay-dependent
-
Product Specifications

Species Reactivity

Pig

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

BL1H8

Immunogen

Porcine alveolar macrophages

Conjugate

FITC FITC FITC

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

0.1 mg/mL

Purification

Protein G

Storage buffer

PBS with 1% BSA

Contains

0.09% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2745234

Product Specific Information

Mouse anti Pig CD11a, clone BL1H8 recognizes porcine CD11a, binding to a conformational epitope on the alpha chain of LFA-1 (CD11a/CD18), expressed on a range of cells including peripheral blood lymphocytes, monocytes, granulocytes and alveolar macrophages.

Target Information

CD11a (LFA-1 alpha, ITGAL) together with CD18 constitute leukocyte function-associated antigen 1 (LFA-1), the alphaLbeta2 integrin. CD11a is implicated in activation of LFA-1 complex. CD11a plays a central role in leukocyte intercellular adhesion through interactions with its ligands, ICAMs 1-3 (intercellular adhesion molecules 1 through 3), and also functions in lymphocyte costimulatory signaling. CD11a is expressed on the plasma membrane of leukocytes in a low-affinity conformation. Cell stimulation by chemokines or other signals leads to induction the high-affinity conformation, which supports tight binding of CD11a to its ligands, the intercellular adhesion molecules ICAM-1, -2, -3. CD11a is thus involved in interaction of various immune cells and in their tissue-specific settlement, but participates also in control of cell differentiation and proliferation and of T-cell effector functions. Blocking of CD11a function by specific antibodies or small molecules has become an important therapeutic approach in treatment of multiple inflammatory diseases. For example, humanized anti- CD11a antibody Efalizumab (Raptiva) is being used to interfere with T cell migration to sites of inflammation, binding of cholesterol-lowering drug simvastatin to CD11a allosteric site leads to immunomodulation and increase in lymphocytic cholinergic activity. Two transcript variants encoding different isoforms of CD11a have been found. Diseases associated with CD11a dysfunction include benign cephalic histiocytosis and leukocyte adhesion deficiency.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: antigen CD11A p180; integrin alpha-L; integrin alphaL

View more View less

Entrez Gene ID: (Pig) 733681

View more View less

It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0