Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CSF1R (CD115) Antibodies

Proteintech

CD115 (c-fms) Monoclonal Antibody (AFS98), CoraLite® Plus 647

View all (80) CSF1R (CD115) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CD115 (c-fms) Monoclonal Antibody (AFS98), CoraLite® Plus 647

  • Antibody Testing Data (1)
CD115 (c-fms) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
CD115 (c-fms) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CD115 (c-fms) Antibody (CL647-65095100T) in Flow

1X10^6 Balb/c Mouse peritoneal macrophages were surface stained with 5 µL CoraLite 647-conjugated Anti-Mouse CD115 (c-fms) (Product # CL647-65095100T), Clone: AFS98) (blue) or CoraLite 647-conjugated Rat IgG2a isotype control (black). Cells were not fixed. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CD115 (c-fms) Antibody in Flow Cytometry (Flow)

Product Details

CL647-65095100T

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

5 µL/1x10^6 cells in a 100 µL suspension
-
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Rat / IgG2a, kappa

Class

Monoclonal

Type

Antibody

Clone

AFS98

Conjugate

CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647
  • Unconjugated
  • APC 
  • CoraLite Plus 488
  • FITC
  • PE 
  • Request custom conjugation

Excitation/Emission Max

654/674 nm

Form

Liquid

Concentration

5 µL/Test

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 0.5% BSA

Contains

0.1% sodium azide

Storage conditions

4°C, store in dark

Shipping conditions

Wet ice

Target Information

CSF1R (FMS) is involved in the production, differentiation, and function of macrophages, and is implicated in promyelocytic leukemias. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. CSF1R is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in the gene encoding CSF1R have been associated with a predisposition to myeloid malignancy.Tyrosine-protein kinase that acts as cell-surface receptor for CSF1 and IL34 and plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. Promotes the release of proinflammatory chemokines in response to IL34 and CSF1, and plays an important role in innate immunity and in inflammatory processes. The first intron of the CSF1R gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. Mutations in the CSF1R gene have been associated with a predisposition to myeloid malignancy.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: c-fms; CD115; CSF-1 receptor; CSF-1-R; CSFIR; EC 2.7.10.1; kinase CSFR; Macrophage colony-stimulating factor 1 receptor; OTTHUMP00000224093; Proto-oncogene c-Fms; proto-oncogene fms

View more View less

Gene Aliases: AI323359; CD115; CSF-1R; Csf1r; Csfmr; Fim-2; Fms; M-CSF-R; M-CSFR

View more View less

UniProt ID: (Mouse) P09581

View more View less

Entrez Gene ID: (Mouse) 12978

View more View less

Function(s)
nucleotide binding protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity macrophage colony-stimulating factor receptor activity protein binding ATP binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups protein phosphatase binding cytokine binding protein homodimerization activity
Process(es)
positive regulation of protein phosphorylation immune system process protein phosphorylation inflammatory response transmembrane receptor protein tyrosine kinase signaling pathway cell proliferation positive regulation of cell proliferation regulation of cell shape phosphorylation peptidyl-tyrosine phosphorylation cytokine-mediated signaling pathway hemopoiesis osteoclast differentiation positive regulation of cell migration ruffle organization cellular response to macrophage colony-stimulating factor stimulus macrophage colony-stimulating factor signaling pathway positive regulation of tyrosine phosphorylation of Stat3 protein innate immune response regulation of bone resorption cell-cell junction maintenance positive regulation of osteoclast differentiation phosphatidylinositol metabolic process protein autophosphorylation phosphatidylinositol-mediated signaling positive regulation of protein tyrosine kinase activity positive regulation of ERK1 and ERK2 cascade cellular response to cytokine stimulus positive regulation of protein serine/threonine kinase activity positive regulation of chemokine secretion positive regulation of cell motility regulation of actin cytoskeleton reorganization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header