Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CAP1 Antibodies

Proteintech

CAP1 Monoclonal Antibody (2H3A10), CoraLite® Plus 488

View all (17) CAP1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite CAP1 Monoclonal Antibody (2H3A10), CoraLite® Plus 488

  • Antibody Testing Data (3)
CAP1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
CAP1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

CAP1 Antibody (CL488-68207) in ICC/IF

Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 488 CAP1 antibody (CL488-68207, Clone: 2H3A10 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

CAP1 Antibody in Immunocytochemistry (ICC/IF)
CAP1 Antibody in Immunocytochemistry (ICC/IF)
CAP1 Antibody in Flow Cytometry (Flow)
CAP1 Monoclonal Antibody (2H3A10), CoraLite® Plus 488

Product Details

CL488-68207

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

1:50-1:500
-

Flow Cytometry (Flow)

0.4 µg/1x10^6 cell suspension
-
Product Specifications

Species Reactivity

Human, Mouse, Pig, Rabbit, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

2H3A10

Immunogen

Recombinant protein
View immunogen

Conjugate

CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
  • Unconjugated
  • Request custom conjugation

Excitation/Emission Max

493/522 nm

Form

Liquid

Purification

Protein G

Storage buffer

PBS with 50% glycerol, 0.5% BSA

Contains

0.05% ProClin 300

Storage conditions

-20°C, store in dark

Shipping conditions

Wet ice

Product Specific Information

Immunogen sequence: SKLFNHLSAV SESIQALGWV AMAPKPGPYV KEMNDAAMFY TNRVLKEYKD VDKKHVDWVK AYLSIWTELQ AYIKEFHTTG LAWSKTGPVA KELSGLPSGP SAGSGPPPPP PGPPPPPVST SSGSDESASR SALFAQINQG ESITHALKHV SDDMKTHKNP ALKAQSGPVR SGPKPFSAPK PQTSPSPKRA TKKEPAVLEL EGKKWRVENQ ENVSNLVIED TELKQVAYIY KCVNTTLQIK GKINSITVDN CKKLGLVFDD VVGIVEIINS KDVKVQVMGK VPTISINKTD GCHAYLSKNS LDCEIVSAKS SEMNVLIPTE GGDFNEFPVP EQFKTLWNGQ KLVTTVTEIA G

Target Information

Adenylyl cyclase-associated protein 1 is an enzyme that in humans is encoded by the CAP1 gene. This gene is mapped to 1p34. 2. The protein encoded by this gene is related to the S. cerevisiae CAP protein, which is involved in the cyclic AMP pathway. And the human protein is able to interact with other molecules of the same protein, as well as with CAP2 and actin. Alternatively spliced transcript variants have been identified.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: adenylyl cyclase-associated CAP protein, yeast homolog 1; adenylyl cyclase-associated protein; Adenylyl cyclase-associated protein 1; CAP 1; CAP, adenylate cyclase-associated protein 1; CAP, adenylate cyclase-associated protein 1 (yeast); cyclase-associated protein homologue; RP11-115D7.1; testis secretory sperm-binding protein Li 218p

View more View less

Gene Aliases: CAP; CAP1; CAP1-PEN; Mch1

View more View less

UniProt ID: (Human) Q01518, (Rat) Q08163, (Mouse) P40124

View more View less

Entrez Gene ID: (Rabbit) 100340076, (Human) 10487, (Rat) 64185, (Mouse) 12331

View more View less

Function(s)
actin binding adenylate cyclase binding molecular_function protein binding
Process(es)
cell morphogenesis ameboidal-type cell migration receptor-mediated endocytosis biological_process actin cytoskeleton organization establishment or maintenance of cell polarity signal transduction activation of adenylate cyclase activity actin polymerization or depolymerization cytoskeleton organization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0