Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • C9orf41 Antibodies

Proteintech

C9orf41 Polyclonal Antibody

View all (15) C9orf41 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite C9orf41 Polyclonal Antibody

  • Antibody Testing Data (4)
C9orf41 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
C9orf41 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

C9orf41 Antibody (24068-1-AP) in WB

HepG2 cells were subjected to SDS PAGE followed by western blot with 24068-1-AP (C9orf41 antibody) at dilution of 1:1000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

C9orf41 Antibody in Western Blot (WB)
C9orf41 Antibody in Western Blot (WB)
C9orf41 Antibody in Western Blot (WB)
C9orf41 Antibody in Western Blot (WB)
C9orf41 Polyclonal Antibody

Product Details

24068-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

C9orf41 Fusion Protein Ag21268 (61-409 aa encoded by BC034033)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.27 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: LEREHFWKI INAFRYYGTS MHERVNRTER QFRSLPANQQ KLLPQFLLHL DKIRKCIDHN QEILLTIVND CIHMFENKEY GEDGNGKIMP ASTFDMDKLK STLKQFVRDW SETGKAERDA CYQPIIKEIL KNFPKERWDP SKVNILVPGA GLGRLAWEIA MLGYACQGNE WSFFMLFSSN FVLNRCSEIN KYKLYPWIHQ FSNNRRSADQ IRPIFFPDVD PHSLPPGSNF SMTAGDFQEI YSECNTWDCI ATCFFIDTAH NVIDYIDTIW KILKPGGIWI NLGPLLYHFE NLANELSIEL SYEDIKNVVL QYGFKVEVEK ESVLSTYTVN DLSMMKYYYE CVLFVVRKPQ (61-409 aa encoded by BC034033)

Target Information

N-methyltransferase that mediates the formation of anserine (beta-alanyl-N(Pi)-methyl-L-histidine) from carnosine. Anserine, a methylated derivative of carnosine (beta-alanyl-L-histidine), is an abundant constituent of vertebrate skeletal muscles. Also methylates other L-histidine-containing di- and tripeptides such as Gly-Gly-His, Gly-His and homocarnosine (GABA-His).

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Carnosine N-methyltransferase; UPF0586 protein C9orf41

View more View less

Gene Aliases: C9orf41; CARNMT1; UPF0586

View more View less

UniProt ID: (Human) Q8N4J0

View more View less

Entrez Gene ID: (Human) 138199

View more View less

Function(s)
carnosine N-methyltransferase activity
Process(es)
methylation carnosine metabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0