Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • C20orf43 Antibodies

OriGene

C20orf43 Monoclonal Antibody (OTI1E8), TrueMAB™

View all (13) C20orf43 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite C20orf43 Monoclonal Antibody (OTI1E8), TrueMAB™

  • Antibody Testing Data (7)
C20orf43 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
C20orf43 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 7

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

C20orf43 Antibody (TA502375) in ICC/IF

Anti-C20orf43 mouse monoclonal antibody (TA502375) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY C20orf43(RC201652). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

C20orf43 Antibody in Immunocytochemistry (ICC/IF)
C20orf43 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
C20orf43 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
C20orf43 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
C20orf43 Antibody in Western Blot (WB)
C20orf43 Antibody in Western Blot (WB)
C20orf43 Antibody in Flow Cytometry (Flow)
C20orf43 Monoclonal Antibody (OTI1E8), TrueMAB™

Product Details

TA502375

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-

Immunocytochemistry (ICC/IF)

1:100
-

Flow Cytometry (Flow)

1:100
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

OTI1E8

Immunogen

Full length human recombinant protein of human C20orf43 produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.58 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

C20orf43 protein function remains unknown.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Replication termination factor 2; Replication termination factor 2 domain-containing protein 1; RTF2; UPF0549 protein C20orf43

View more View less

Gene Aliases: AD-007; C20orf43; CDA05; CDAO5; HSPC164; HSPC169; RTF2; RTFDC1; SHUJUN-3

View more View less

UniProt ID: (Human) Q9BY42

View more View less

Entrez Gene ID: (Human) 51507

View more View less

Function(s)
molecular_function
Process(es)
biological_process mitotic DNA replication termination
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0