Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • C1QL4 Antibodies

Invitrogen

C1QL4 Polyclonal Antibody

View all (2) C1QL4 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite C1QL4 Polyclonal Antibody

C1QL4 Polyclonal Antibody

Product Details

PA5-68158

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:3,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthesized peptide derived from human C1QL4.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2691730

Product Specific Information

Predicted to react with Mouse and Rat samples.

Target Information

May regulate the number of excitatory synapses that are formed on hippocampus neurons. Has no effect on inhibitory synapses. May inhibit adipocyte differentiation at an early stage of the process.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: C1q and tumor necrosis factor-related protein 11; C1q/TNF-related protein 11; C1qTNF11; Complement C1q-like protein 4; complement component 1, q subcomponent like 4; complement component 1, q subcomponent-like 4

View more View less

Gene Aliases: C1QL4; C1QTNF11; CTRP11

View more View less

UniProt ID: (Human) Q86Z23, (Mouse) Q4ZJM9

View more View less

Entrez Gene ID: (Human) 338761, (Mouse) 239659

View more View less

Function(s)
identical protein binding protein binding
Process(es)
negative regulation of fat cell differentiation negative regulation of fibroblast proliferation negative regulation of ERK1 and ERK2 cascade
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0