Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • C1QA Antibodies

NeoBiotechnologies

C1QA/Complement C1q A-Chain Monoclonal Antibody (C1QA, 2783)

View all (28) C1QA antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite C1QA/Complement C1q A-Chain Monoclonal Antibody (C1QA, 2783)

C1QA/Complement C1q A-Chain Monoclonal Antibody (C1QA, 2783)
  • Antibody Testing Data (4)
C1QA/Complement C1q A-Chain Antibody in Western Blot (WB)
Group 53 Created with Sketch.
C1QA/Complement C1q A-Chain Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

C1QA/Complement C1q A-Chain Antibody (712-MSM1-P1ABX) in WB

Western Blot analysis of human spleen, PC3 and liver lysates using C1QA, Complement C1q A-Chain Monoclonal Antibody (Product # 712-MSM1-P1ABX). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

C1QA/Complement C1q A-Chain Antibody in Western Blot (WB)
C1QA/Complement C1q A-Chain Antibody in Western Blot (WB)
C1QA/Complement C1q A-Chain Antibody in Peptide array (ARRAY)
C1QA/Complement C1q A-Chain Antibody in SDS-PAGE (SDS-PAGE)

Product Details

712-MSM1-P1ABX

Applications
Tested Dilution
Publications

Western Blot (WB)

1-2 µg/mL
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b, kappa

Class

Monoclonal

Type

Antibody

Clone

C1QA, 2783

Immunogen

Recombinant fragment (around aa 104-237) of human C1QA protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Antibody is tested in direct ELISA.

Target Information

C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: complement C1q chain A; Complement C1q subcomponent subunit A; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component C1q, A chain

View more View less

Gene Aliases: C1QA

View more View less

UniProt ID: (Human) P02745

View more View less

Entrez Gene ID: (Human) 712

View more View less

Function(s)
serine-type endopeptidase activity protein binding
Process(es)
proteolysis complement activation complement activation, classical pathway cell-cell signaling response to iron ion innate immune response
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5f5c96ff6c-st5hm:80/100.66.129.203:80.
git-commit: 945c3ce0b94337c0c8e9574aed9898c23d0d1109
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0