Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • C1 inhibitor Antibodies

OriGene

C1 inhibitor Polyclonal Antibody

View all (24) C1 inhibitor antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite C1 inhibitor Polyclonal Antibody

  • Antibody Testing Data (1)
C1 inhibitor Antibody in ELISA (ELISA)
Group 53 Created with Sketch.
C1 inhibitor Antibody in ELISA (ELISA)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

C1 inhibitor Antibody (TA590004) in ELISA

ELISA: C1 Inactivator Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

C1 inhibitor Antibody in ELISA (ELISA)
C1 inhibitor Polyclonal Antibody

Product Details

TA590004

Applications
Tested Dilution
Publications

ELISA (ELISA)

1:100-1:2,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

DNA immunization. This antibody is specific for the N Terminus Region of the target protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.70146 mg/mL

Purification

Protein A/G

Storage buffer

PBS

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

C1-inhibitor (C1INH) functions as a regulator of the activation of the classical complement pathway and of the contact activation system of kinin generation and coagulation. Its primary biologically relevant target proteinases are C1r, C1s, coagulation factors XIa and XIIa, and plasma kallikrein. C1-inhibitor consists of one polypeptide chain and has a molecular mass of ~104 kDa. Serum concentration has been determined to 180 µg/mL.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: C1 esterase inhibitor; C1 Inh; C1-inhibiting factor; complement component 1 inhibitor; esterase inhibitor; IC1; Plasma protease C1 inhibitor; serine/cysteine proteinase inhibitor clade G member 1; Serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1

View more View less

Gene Aliases: C1IN; C1INH; C1NH; HAE1; HAE2; SERPING1

View more View less

UniProt ID: (Human) P05155

View more View less

Entrez Gene ID: (Human) 710

View more View less

Function(s)
serine-type endopeptidase inhibitor activity protein binding
Process(es)
negative regulation of complement activation, lectin pathway platelet degranulation complement activation, classical pathway aging blood coagulation, intrinsic pathway blood circulation negative regulation of endopeptidase activity fibrinolysis innate immune response
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0