Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Aurora B Antibodies

NeoBiotechnologies

Aurora B (Proliferation Marker) Monoclonal Antibody (rAURKB/1592)

View all (46) Aurora B antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Aurora B (Proliferation Marker) Monoclonal Antibody (rAURKB/1592)

  • Antibody Testing Data (4)
Aurora B (Proliferation Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Aurora B (Proliferation Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Aurora B (Proliferation Marker) Antibody (9212-MSM7-P1ABX) in IHC (P)

Formalin-fixed, paraffin-embedded human placenta stained with Aurora B Recombinant Mouse Monoclonal Antibody (rAURKB/1592). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Aurora B (Proliferation Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Aurora B (Proliferation Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Aurora B (Proliferation Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Aurora B (Proliferation Marker) Antibody in SDS-PAGE (SDS-PAGE)
Aurora B (Proliferation Marker) Monoclonal Antibody (rAURKB/1592)

Product Details

9212-MSM7-P1ABX

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

rAURKB/1592

Immunogen

Recombinant fragment (around aa 89-251) of human Aurora B protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

The Aurora kinases associate with microtubules during chromosome movement and segregation. AURKB (Aurora B) localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers. This complex functions in chromosome alignment and separation, histone modification, and cytokinesis. CPC also includes three non-enzymatic subunits known as the inner centromere protein (INCENP), survivin, and borealin, which determine the activity, localization, stability, and possibly even the substrate specificity of AurB. Two CPCs exist during mitosis: one containing all four members and another consisting of INCENP and AurB. Quaternary CPC functions during chromosome alignment and cytokinesis, whereas the INCENP-AurB complex may be responsible for modifying histone H3. AurB is overexpressed in a number of tumours, including primary breast and colon tumour samples. Its expression in tumours seems to be linked with Aurora-A kinase, suggesting a feedback mechanism between the two proteins.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: AIM-1; ARK-2; Aurora 1; Aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; Aurora- and IPL1-like midbody-associated protein 1; aurora-1; aurora-B; aurora-related kinase 2; Aurora/IPL1-related kinase 2; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase 12; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase aurora-B; STK-1

View more View less

Gene Aliases: AIK2; AIM-1; AIM1; AIRK2; ARK2; AurB; AURKB; aurkb-sv1; aurkb-sv2; IPL1; PPP1R48; STK1; STK12; STK5

View more View less

UniProt ID: (Human) Q96GD4

View more View less

Entrez Gene ID: (Human) 9212

View more View less

Function(s)
protein serine/threonine kinase activity protein serine/threonine/tyrosine kinase activity protein binding ATP binding histone serine kinase activity metal ion binding
Process(es)
negative regulation of transcription from RNA polymerase II promoter negative regulation of B cell apoptotic process protein phosphorylation spindle organization sister chromatid cohesion aging cell proliferation attachment of spindle microtubules to kinetochore abscission histone modification protein sumoylation anaphase-promoting complex-dependent catabolic process spindle checkpoint negative regulation of protein binding positive regulation of telomere maintenance via telomerase negative regulation of cytokinesis positive regulation of cytokinesis protein localization to kinetochore cellular response to UV cleavage furrow formation protein ubiquitination involved in ubiquitin-dependent protein catabolic process histone H3-S28 phosphorylation protein autophosphorylation mitotic spindle midzone assembly positive regulation of telomerase activity regulation of chromosome segregation regulation of signal transduction by p53 class mediator positive regulation of telomere capping
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0