Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ACE Antibodies

NeoBiotechnologies

Angiotensin I Converting Enzyme (ACE)/CD143 Monoclonal Antibody (ACE/3762)

View all (40) ACE antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Angiotensin I Converting Enzyme (ACE)/CD143 Monoclonal Antibody (ACE/3762)

Angiotensin I Converting Enzyme (ACE)/CD143 Monoclonal Antibody (ACE/3762)
  • Antibody Testing Data (3)
Angiotensin I Converting Enzyme (ACE)/CD143 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Angiotensin I Converting Enzyme (ACE)/CD143 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Angiotensin I Converting Enzyme (ACE)/CD143 Antibody (1636-MSM2-P1ABX) in IHC (P)

Formalin-fixed, paraffin-embedded human colon stained with ACE/CD143 Mouse Monoclonal Antibody (ACE/3762). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Angiotensin I Converting Enzyme (ACE)/CD143 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Angiotensin I Converting Enzyme (ACE)/CD143 Antibody in Western Blot (WB)
Angiotensin I Converting Enzyme (ACE)/CD143 Antibody in SDS-PAGE (SDS-PAGE)

Product Details

1636-MSM2-P1ABX

Applications
Tested Dilution
Publications

Western Blot (WB)

1-2 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2c, kappa

Class

Monoclonal

Type

Antibody

Clone

ACE/3762

Immunogen

Recombinant fragment (around aa 652-788) of human ACE/CD143 protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Target Information

This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287 bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes-the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: ACE; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; Angiotensin-converting enzyme; carboxycathepsin; CD143; CD143 antigen; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase I; Kininase II; MGC26566; peptidase P; testicular ECA

View more View less

Gene Aliases: ACE; ACE1; CD143; DCP; DCP1; ICH; MVCD3

View more View less

UniProt ID: (Human) P12821

View more View less

Entrez Gene ID: (Human) 1636

View more View less

Function(s)
endopeptidase activity carboxypeptidase activity drug binding metallopeptidase activity exopeptidase activity tripeptidyl-peptidase activity peptidyl-dipeptidase activity zinc ion binding chloride ion binding mitogen-activated protein kinase kinase binding bradykinin receptor binding mitogen-activated protein kinase binding metallodipeptidase activity
Process(es)
kidney development blood vessel remodeling angiotensin maturation angiotensin catabolic process in blood regulation of renal output by angiotensin neutrophil mediated immunity antigen processing and presentation of peptide antigen via MHC class I regulation of systemic arterial blood pressure by renin-angiotensin spermatogenesis regulation of blood pressure regulation of smooth muscle cell migration regulation of vasoconstriction mononuclear cell proliferation regulation of vasodilation hormone catabolic process peptide catabolic process beta-amyloid metabolic process arachidonic acid secretion heart contraction regulation of angiotensin metabolic process hematopoietic stem cell differentiation positive regulation of protein tyrosine kinase activity cell proliferation in bone marrow positive regulation of peptidyl-tyrosine autophosphorylation regulation of hematopoietic stem cell proliferation negative regulation of gap junction assembly positive regulation of peptidyl-cysteine S-nitrosylation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-564b8c87b9-4vhxl:80/100.66.128.26:80.
git-commit: e423ab1ff6713b59ee60fc1fa9b2d3517c9165b4
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0