Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • SNAP alpha Antibodies

Proteintech

Alpha SNAP Monoclonal Antibody (1E2A2), CoraLite® Plus 488

View all (15) SNAP alpha antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Alpha SNAP Monoclonal Antibody (1E2A2), CoraLite® Plus 488

  • Antibody Testing Data (2)
Alpha SNAP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Alpha SNAP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Alpha SNAP Antibody (CL48867323100UL) in IHC (P)

Immunofluorescent analysis of (4% PFA) fixed Mouse brain tissue using CoraLite 488 Alpha SNAP antibody (Product # CL48867323100UL), Clone: 1E2A2 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Alpha SNAP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Alpha SNAP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Alpha SNAP Monoclonal Antibody (1E2A2), CoraLite® Plus 488

Product Details

CL48867323100UL

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:500
-
Product Specifications

Species Reactivity

Human, Mouse, Pig, Rat

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

1E2A2

Conjugate

CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
  • Unconjugated
  • CoraLite 594
  • Request custom conjugation

Excitation/Emission Max

493/522 nm

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 50% glycerol, 0.5% BSA

Contains

0.05% ProClin 300

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles, store in dark

Shipping conditions

Wet ice

Target Information

SNAPs (soluble NSF attachment proteins), acting in concert with SNAREs (SNAP receptors) and the N-ethylmaleimide-sensitive fusion protein (NSF) are required for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-to-endosome fusion and transcytotic vesicles-to-plasma membrane transport. Vesicle-to-target membrane docking (initial contact) occurs when the vesicle SNARE binds to its cognate target membrane SNARE. a-SNAP (or b-SNAP in brain) then binds to this docking complex and mediates the binding of NSF and thus the formation of a 20S fusion particle. It is thought that, once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: alpha snap; alpha soluble NSF attachment protein; alpha-SNAP; Alpha-soluble NSF attachment protein; hydrocephaly with hop gait; N-ethylmaleimide sensitive fusion protein attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein, alpha; SNAP-alpha

View more View less

Gene Aliases: 1500039N14Rik; a-SNAP; alpha-SNAP; AW209189; hyh; NAPA; RA81; Snap; SNAPA; SNARE

View more View less

UniProt ID: (Human) P54920, (Rat) P54921, (Mouse) Q9DB05

View more View less

Entrez Gene ID: (Pig) 100515953, (Human) 8775, (Rat) 140673, (Mouse) 108124

View more View less

Function(s)
soluble NSF attachment protein activity protein binding syntaxin binding SNARE binding
Process(es)
intracellular protein transport brain development regulation of synaptic vesicle priming neuron differentiation synaptic transmission, glutamatergic SNARE complex disassembly apical protein localization membrane fusion transport protein transport vesicle-mediated transport ER to Golgi vesicle-mediated transport retrograde vesicle-mediated transport, Golgi to ER intra-Golgi vesicle-mediated transport COPII vesicle coating
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0