Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Secondary Antibodies ›
  • Alpaca anti-Mouse IgG3 Secondary Antibodies

Chromotek

Alpaca anti-Mouse IgG3 VHH, Nano-Secondaries™ Recombinant Secondary Antibody, Alexa Fluor™ 647

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Create Citation
Questions & Answers
Datasheet
Create Citation
Questions & Answers

Cite Alpaca anti-Mouse IgG3 VHH, Nano-Secondaries™ Recombinant Secondary Antibody, Alexa Fluor™ 647

  • Antibody Testing Data (3)
Mouse IgG3 VHH Secondary Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Mouse IgG3 VHH Secondary Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Mouse IgG3 VHH Secondary Antibody (SMS3AF647-1-100) in ICC/IF

Multiplexed immunostaining of HeLa cells with two alpaca anti-Mouse Nano-Secondaries and one conventional secondary antibody. Green: Mouse IgG3 anti-Lamin + alpaca anti-Mouse IgG3 VHH Alexa Fluor® 488. Red: Mouse IgG1 anti-Tubulin + alpaca anti-Mouse gG1 VHH Alexa Fluor® 568. Magenta: rabbit anti-Ki67 + conventional polyclonal secondary anti-rabbit-AF647. Scale bar, 10 μm. Images were recorded at the Core F... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Mouse IgG3 VHH Secondary Antibody in Immunocytochemistry (ICC/IF)
Mouse IgG3 VHH Secondary Antibody in Immunocytochemistry (ICC/IF)
Mouse IgG3 VHH Secondary Antibody in Immunocytochemistry (ICC/IF)

Product Details

SMS3AF647-1-100

Applications
Tested Dilution

Western Blot (WB)

1:1,000

Immunocytochemistry (ICC/IF)

1:1,000
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Alpaca

Expression System

E. coli

Class

Recombinant Monoclonal

Type

Secondary Antibody

Clone

CTK0107

Immunogen

mouse IgG3

Conjugate

Alexa Fluor™ 647 Alexa Fluor™ 647 Alexa Fluor™ 647

Excitation/Emission Max

650/671 nm View spectra spectra

Form

Liquid

Concentration

0.5 mg/mL

Purification

Affinity chromatography

Storage buffer

25mM TAPS, pH 8.5, with 5mM EDTA, 500mM NaCl

Contains

0.09% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Target

IgG3

Antibody Form

VHH

Product Specific Information

Anti-mouse IgG3 Nanobody for detection of mouse IgG3 in Immunofluorescence and Western Blot.

Affinity: CTK0107: KD = 5.7 nM.

Target Information

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

View product citations for antibody SMS3AF647-1-100 on CiteAb

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0