Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • AOX1 Antibodies

Proteintech

Aldehyde oxidase Polyclonal Antibody

10 References
View all (10) AOX1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Aldehyde oxidase Polyclonal Antibody

  • Antibody Testing Data (9)
Aldehyde oxidase Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Aldehyde oxidase Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 9

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Aldehyde oxidase Antibody (19495-1-AP) in ICC/IF

Immunofluorescent analysis of HepG2 cells, using AOX1 antibody 19495-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). Blue pseudocolor = DAPI (fluorescent DNA dye). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Aldehyde oxidase Antibody in Immunocytochemistry (ICC/IF)
Aldehyde oxidase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Aldehyde oxidase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Aldehyde oxidase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Aldehyde oxidase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Aldehyde oxidase Antibody in Western Blot (WB)
Aldehyde oxidase Antibody in Western Blot (WB)
Aldehyde oxidase Antibody in Western Blot (WB)
Aldehyde oxidase Antibody in Immunoprecipitation (IP)

Product Details

19495-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
View 7 publications 7 publications

Immunohistochemistry (IHC)

-
View 2 publications 2 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:20-1:200
-

Immunocytochemistry (ICC/IF)

1:10-1:100
View 1 publication 1 publication

Immunoprecipitation (IP)

0.5-4.0 µg
-
Product Specifications

Species Reactivity

Dog, Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Peptide

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.43 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Target Information

AOX1 (Aldehyde oxidase 1) is a 1338 amino acid cytoplasmic protein that catalyzes the oxidation of a variety of aldehydes, leading to the production of hydrogen peroxide. Under certain conditions, AOX1 can catalyze the formation of the superoxide free radical. Defects in oxygen radical metabolism have been linked to the pathogenesis of amyotrophic lateral sclerosis (ALS), an autosomal dominant neurodegenerative disorder characterized by the death of motor neurons in the spinal cord, brain and brainstem. Significantly, AOX1 is highly expressed in the ventral horn of the spinal cord and the gene that encodes AOX1 is located in a chromosomal region that is frequently found to be implicated in ALS2. This evidence suggests that AOX1 is a candidate gene for ALS2.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: Aldehyde oxidase; aldehyde oxidase (female form); Aldehyde oxidase 1; aldehyde oxidase 2; Azaheterocycle hydroxylase; Azaheterocycle hydroxylase 1; Retinal oxidase

View more View less

Gene Aliases: AI196512; AI255253; AO; AOH1; Aox-1; Aox-2; AOX1; Aox2; Moro; Ro

View more View less

UniProt ID: (Human) Q06278, (Mouse) O54754, (Rat) Q9Z0U5

View more View less

Entrez Gene ID: (Human) 316, (Mouse) 11761, (Rat) 54349

View more View less

Function(s)
aldehyde oxidase activity xanthine dehydrogenase activity iron ion binding electron carrier activity oxidoreductase activity, acting on CH-OH group of donors molybdopterin cofactor binding flavin adenine dinucleotide binding NAD binding 2 iron, 2 sulfur cluster binding catalytic activity UDP-N-acetylmuramate dehydrogenase activity oxidoreductase activity metal ion binding iron-sulfur cluster binding
Process(es)
oxidation-reduction process inflammatory response xanthine catabolic process vitamin B6 metabolic process reactive oxygen species metabolic process xenobiotic metabolic process drug metabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0