Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Adenylate kinases are involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate groups among adenine nucleotides. Three isozymes of adenylate kinase, namely 1, 2, and 3, have been identified in vertebrates; this gene encodes isozyme 2. Expression of these isozymes is tissue-specific and developmentally regulated. Isozyme 2 is localized in the mitochondrial intermembrane space and may play a role in apoptosis. Two transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate kinase 2, mitochondrial; adenylate kinase 2, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03168}; adenylate kinase isoenzyme 2, mitochondrial; Adenylate monophosphate kinase; adenylate monophosphate kinase {ECO:0000255|HAMAP-Rule:MF_03168}; AK 2; AK 2 {ECO:0000255|HAMAP-Rule:MF_03168}; ATP-AMP transphosphorylase 2; ATP-AMP transphosphorylase 2 {ECO:0000255|HAMAP-Rule:MF_03168}; ATP:AMP phosphotransferase; ATP:AMP phosphotransferase {ECO:0000255|HAMAP-Rule:MF_03168}; testis secretory sperm-binding protein Li 220n
Gene Aliases: ADK2; Ak-2; AK2; D4Ertd220e
UniProt ID: (Human) P54819, (Rat) P29410, (Mouse) Q9WTP6
Entrez Gene ID: (Human) 204, (Rat) 24184, (Mouse) 11637
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support