Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ARMX2 Antibodies

Invitrogen

ARMX2 Polyclonal Antibody

View all (8) ARMX2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite ARMX2 Polyclonal Antibody

ARMX2 Polyclonal Antibody

Product Details

PA5-50912

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

Immunohistochemistry (IHC)

1:100-1:300
-

Immunocytochemistry (ICC/IF)

1:200-1:1,000
-

ELISA (ELISA)

1:20,000
-
Product Specifications

Species Reactivity

Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthesized peptide derived from the Internal region of human ARMCX2 (aa 321-370).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol, 0.5% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2636360

Product Specific Information

ARMCX2 Polyclonal Antibody detects endogenous levels of ARMCX2 protein.

Target Information

The armadillo (ARM) repeat family of proteins are related to the Drosophila melanogaster armadillo protein, a protein essential for wingless signal transduction. ARM proteins are involved in a variety of processes such as cell migration, cell proliferation, tissue maintenance and tumorigenesis, and they also function in signal transduction and the maintenance of overall cell structure. ARMCX2 (armadillo repeat containing, X-linked 2), also known as ALEX2, is a 632 amino acid single-pass membrane protein that contains three ARM repeats and is highly expressed in testis, brain, ovary, heart, colon, spleen and prostate, where it is thought to play a role in tumor suppression. The gene encoding ARMCX2 maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1, 000 genes.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: ALEX2 protein; armadillo repeat-containing X-linked protein 2

View more View less

Entrez Gene ID: (Rat) 367903

View more View less

It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0