Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Broad-specificity nucleoside monophosphate (NMP) kinase that catalyzes the reversible transfer of the terminal phosphate group between nucleoside triphosphates and monophosphates. AMP and dAMP are the preferred substrates, but CMP and dCMP are also good substrates. IMP is phosphorylated to a much lesser extent. All nucleoside triphosphates ATP, GTP, UTP, CTP, dATP, dCTP, dGTP, and TTP are accepted as phosphate donors. CTP is the best phosphate donor, followed by UTP, ATP, GTP and dCTP. May have a role in nuclear energy homeostasis. Has also ATPase activity. May be involved in regulation of Cajal body (CB) formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate kinase isoenzyme 6; adenylate kinase isoenzyme 6 {ECO:0000255|HAMAP-Rule:MF_03173}; Adrenal gland protein AD-004; AK/ATPase; AK/ATPase {ECO:0000255|HAMAP-Rule:MF_03173}; AK6; AK6 {ECO:0000255|HAMAP-Rule:MF_03173}; ATP-AMP transphosphorylase 6; coilin interacting nuclear ATPase protein; coilin interacting protein; Coilin-interacting nuclear ATPase protein; coilin-interacting nuclear ATPase protein {ECO:0000255|HAMAP-Rule:MF_03173}; Dual activity adenylate kinase/ATPase; dual activity adenylate kinase/ATPase {ECO:0000255|HAMAP-Rule:MF_03173}; hCINAP
Gene Aliases: 2810046E22Rik; 4921516M08Rik; AD-004; AK/ATPase; AK6; CGI-137; CINAP; CIP; hCINAP
UniProt ID: (Human) Q9Y3D8, (Rat) Q5EB68, (Mouse) Q8VCP8
Entrez Gene ID: (Human) 102157402, (Rat) 102238592, (Mouse) 102216272
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support