Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • AHSP Antibodies

Invitrogen

AHSP Polyclonal Antibody

View all (7) AHSP antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite AHSP Polyclonal Antibody

  • Antibody Testing Data (3)
AHSP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
AHSP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

AHSP Antibody (PA5-118943) in IHC (P)

Immunohistochemistry analysis of AHSP in paraffin-embedded Mouse Stomach tissue. Samples were incubated with polyclonal antibody (Product # PA5-118943) at a dilution of 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

AHSP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
AHSP Antibody in Western Blot (WB)
AHSP Antibody in Western Blot (WB)
AHSP Polyclonal Antibody

Product Details

PA5-118943

Applications
Tested Dilution
Publications

Western Blot (WB)

0.01-3 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

5-30 µg/mL
-
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant protein Alpha-Hemoglobin Stabilizing Protein. The antigen corresponds to amino acid range 2-102 of the target protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5 mg/mL

Purification

Antigen affinity chromatography, Protein A

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2903443

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Alpha-hemoglobin-stabilizing protein; erythroid associated factor; Erythroid differentiation-related factor; Erythroid-associated factor

View more View less

Gene Aliases: Ahsp; EDRF; Eraf

View more View less

UniProt ID: (Mouse) Q9CY02

View more View less

Entrez Gene ID: (Mouse) 170812

View more View less

Function(s)
hemoglobin binding
Process(es)
protein folding hemoglobin metabolic process hemopoiesis erythrocyte differentiation protein stabilization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0