Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ADRM1 Antibodies

Bioss

ADRM1 Polyclonal Antibody

View all (14) ADRM1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite ADRM1 Polyclonal Antibody

ADRM1 Polyclonal Antibody

Product Details

BS-13357R

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:200-1:400
-

Immunohistochemistry (Frozen) (IHC (F))

1:100-1:500
-

Immunocytochemistry (ICC/IF)

1:100-1:500
-

ELISA (ELISA)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic peptide derived from human ADRM1, amino acids 21-120.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

0.01M TBS, pH 7.4, with 1% BSA, 50% glycerol

Contains

0.02% ProClin 300

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

The protein encoded by this gene is an integral plasma membrane protein which promotes cell adhesion. The encoded protein is thought to undergo O-linked glycosylation. Expression of this gene has been shown to be induced by gamma interferon in some cancer cells. Two transcript variants encoding the same protein have been found for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 110 kDa cell membrane glycoprotein; Adhesion-regulating molecule 1; ARM-1; cell membrane glycoprotein, 110000M(r) (surface antigen); glycoprotein 110; Gp110; hRpn13; M(r) 110,000 surface antigen; MGC29536; Proteasomal ubiquitin receptor ADRM1; Proteasome regulatory particle non-ATPase 13; Rpn13 homolog

View more View less

Gene Aliases: 1110063P18Rik; 2510006J17Rik; AA408205; ADRM1; ARM-1; ARM1; AU043535; GP110; Rpn13

View more View less

UniProt ID: (Human) Q16186, (Rat) Q9JMB5, (Mouse) Q9JKV1

View more View less

Entrez Gene ID: (Human) 11047, (Rat) 65138, (Mouse) 56436

View more View less

Function(s)
protease binding protein binding endopeptidase activator activity proteasome binding
Process(es)
ovarian follicle development transcription elongation from RNA polymerase II promoter spermatid development positive regulation of endopeptidase activity regulation of T cell differentiation in thymus follicle-stimulating hormone signaling pathway proteasome assembly oogenesis thymus development Sertoli cell development positive regulation of growth hormone receptor signaling pathway adipose tissue development seminiferous tubule development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0