Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ADCY2 Antibodies

Fabgennix

ADCY2 Monoclonal Antibody (O18.43a), FITC

View all (18) ADCY2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite ADCY2 Monoclonal Antibody (O18.43a), FITC

ADCY2 Monoclonal Antibody (O18.43a), FITC

Product Details

ADCY2-101MAB-FITC

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500
-

ELISA (ELISA)

1:10,000
-

Immunoprecipitation (IP)

1:200
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Mouse / IgG

Class

Monoclonal

Type

Antibody

Clone

O18.43a

Immunogen

Synthetic peptide corresponding to amino acids within region 950-1000 on mouse ADCY2 protein.
View immunogen

Conjugate

FITC FITC FITC

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

0.5-1.5 µg/mL

Purification

Protein G

Storage buffer

proprietary buffer with 30% glycerol

Contains

0.01% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

ADCY2 is a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate(cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 3',5'-cyclic AMP synthetase; AC2; adenylate cyclase 2 (brain); adenylate cyclase II; Adenylate cyclase type 2; Adenylate cyclase type II; Adenylyl cyclase 2; ATP pyrophosphate-lyase 2; type II adenylate cyclase

View more View less

Gene Aliases: AC2; ADCY2; HBAC2; KIAA1060; mKIAA1060

View more View less

UniProt ID: (Human) Q08462, (Rat) P26769

View more View less

Entrez Gene ID: (Human) 108, (Mouse) 210044, (Rat) 81636

View more View less

Function(s)
magnesium ion binding adenylate cyclase activity ATP binding adenylate cyclase binding manganese ion binding protein heterodimerization activity nucleotide binding calcium- and calmodulin-responsive adenylate cyclase activity lyase activity phosphorus-oxygen lyase activity G-protein beta/gamma-subunit complex binding metal ion binding protein binding
Process(es)
cAMP biosynthetic process adenylate cyclase-modulating G-protein coupled receptor signaling pathway activation of adenylate cyclase activity cyclic nucleotide biosynthetic process intracellular signal transduction renal water homeostasis adenylate cyclase-activating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway cAMP-mediated signaling activation of protein kinase A activity cellular response to glucagon stimulus cellular response to forskolin
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0