Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • 4-1BBL (CD137L) Antibodies

Invitrogen

4-1BB Ligand Polyclonal Antibody, Biotin, PeproTech®

View all (44) 4-1BBL (CD137L) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite 4-1BB Ligand Polyclonal Antibody, Biotin, PeproTech®

  • Antibody Testing Data (3)
4-1BB Ligand Antibody in Western Blot (WB)
Group 53 Created with Sketch.
4-1BB Ligand Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

4-1BB Ligand Antibody (500-P169BT-25UG) in WB

Western Blot: To detect h4-1BBL by Western Blot analysis 4-1BB Ligand Polyclonal Antibody, Biotin (Product # 500-P169BT-1MG) can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant h4-1BBL is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

4-1BB Ligand Antibody in Western Blot (WB)
4-1BB Ligand Antibody in Western Blot (WB)
4-1BB Ligand Antibody in ELISA (ELISA)
4-1BB Ligand Polyclonal Antibody, Biotin, PeproTech®

Product Details

500-P169BT-25UG

Applications
Tested Dilution
Publications

Western Blot (WB)

0.1-0.2 µg/mL
-

ELISA (ELISA)

0.25-1.0 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit

Class

Polyclonal

Type

Antibody

Immunogen

E.coli-derived Recombinant Human 4-1BB Ligand
View immunogen

Conjugate

Biotin Biotin Biotin

Form

Lyophilized

Concentration

0.1-1.0 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS

Contains

no preservative

Storage conditions

-20°C

Shipping conditions

Ambient

RRID

AB_2929610

Product Specific Information

AA Sequence of recombinant protein: MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE.

Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human 4-1BB Ligand. Anti-Human 4-1BB Ligand-specific antibody was purified by affinity chromatography and then biotinylated.

Sandwich ELISA: To detect h4-1BBL by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human 4-1BBL (500-P169) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant h4-1BBL.

Western Blot: To detect h4-1BBL by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant h4-1BBL is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

500-P169BT-1MG will be provided as 2 x 500 µg

Target Information

CD137 ligand (CD137L) is a type II membrane protein and part of the TNF superfamily. CD137L is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation. Its receptor, CD137/4-1BB, is found on a variety of cells, including inflamed endothelial cells, where its expression enhances extravasation of CD137L expressing monocytes. Activated T cells also express CD137, and engagement with CD137L enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137L is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but it is lacking on Hodgkin lymphoma and T-cell lymphoma). CD137L is also expressed in carcinoma cell lines, and is thought to be involved in T cell-tumor cell interactions.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 4 1 BB Ligand; 4 1 BBL; 4-1BB ligand; 4-1BBL; 41BB Ligand; 41BBL; CD 137 Ligand; CD 137L; CD137 Ligand; CDw137L; homolog of mouse 4-1BB-L; receptor 4-1BB ligand; TNFRSF9; tumor necrosis factor (ligand) superfamily member 9; tumor necrosis factor (ligand) superfamily, member 9; tumor necrosis factor ligand 5A; Tumor necrosis factor ligand superfamily member 9

View more View less

Gene Aliases: 4-1BB-L; CD137L; TNFSF9; TNLG5A

View more View less

UniProt ID: (Human) P41273

View more View less

Entrez Gene ID: (Human) 8744

View more View less

Function(s)
receptor binding cytokine activity tumor necrosis factor receptor binding tumor necrosis factor receptor superfamily binding
Process(es)
apoptotic process immune response signal transduction cell-cell signaling cell proliferation positive regulation of interferon-gamma production positive regulation of interleukin-12 production positive regulation of interleukin-6 production positive regulation of activated T cell proliferation myeloid dendritic cell differentiation positive regulation of cytotoxic T cell differentiation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0