Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55656. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Plays a role in tyrosine phosphorylation of CBL by linking CBL to the insulin receptor. Required for insulin-stimulated glucose transport. Involved in formation of actin stress fibers and focal adhesions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Cbl associated protein; c-Cbl-associated protein; CAP; DKFZp451C066; DKFZP586P1422; Fas-ligand associated factor 2; FLJ12406; OTTHUMP00000020142; OTTHUMP00000020143; OTTHUMP00000020144; OTTHUMP00000020145; Ponsin; SH3 domain protein 5; SH3P12; Sorbin and SH3 domain-containing protein 1; SORBS1
Gene Aliases: CAP; FLAF2; KIAA0894; KIAA1296; R85FL; SH3D5; SH3P12; SORB1; SORBS1
UniProt ID: (Human) Q9BX66
Entrez Gene ID: (Human) 10580
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support