• ELISA Kits ›
  • TCR V beta 6 ELISA Kits

Invitrogen

TCR V beta 6 Competitive ELISA Kit

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
TCR V beta 6 Competitive ELISA Kit
Group 53 Created with Sketch.
TCR V beta 6 Competitive ELISA Kit
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

TCR V beta 6 Competitive ELISA Kit

ELISA Standard Curve of TCR V beta 6 using TCR V beta 6 Competitive ELISA Kit (Product # EEL148). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

TCR V beta 6 Competitive ELISA Kit
TCR V beta 6 Competitive ELISA Kit (EEL148)
videoThumbnail
►

Product Specifications

Analytical sensitivity

0.94 ng/mL

Assay range

1.56-100 ng/mL

Sample type/volume

Cell Lysate
50 µL
Plasma
50 µL
Serum
50 µL
Tissue Homogenate
50 µL

Hands-on time

1 hr 20 min

Time-to-result

2 hr 30 min

Homogenous (no wash)

No

Interassay CV

<10%

Intraassay CV

<10%

Instrument

Absorbance Microplate Reader

Product size

96 Tests

Contents

Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer

Shipping conditions

Wet ice

Storage

Refer to product documentation for component specific storage temperature.

Protein name

TCR V beta 6

Species (tested)

All

Species (reported)

All

Assay kit format

Competitive ELISA

Detector antibody conjugate

Biotin

Label or dye

HRP

About This Kit

The Vitamin B6 (VB6) ELISA quantitates VB6 in Serum, plasma and other biological fluids.

Principle of the method

The Competitive multispecies ELISA research use only kit is designed to quantitatively measure Vitamin B6 (VB6) independent of species. A VB6 standard is provided to generate a standard curve for the assay and all samples are read off a user generated standard curve. Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to VB6 is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of VB6 in the samples is then determined by comparing the OD of the samples to the standard curve.

Rigorous validation:

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Target information

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases : TCR V beta6, TCRV beta 6, TCRV beta6, Vb6, Vbeta6

Recommended Products

IL-6 Human Matched Antibody Pair

Because you are interested in IL-6 Human ELISA Kit.

SIZE

10 x 96 tests

PRICE

USD 400.00

Cat # CHC1263

guarantee_icon

Performance Guarantee

If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-585975bbdc-jwj8f:80/100.66.129.203:80.
git-commit: a17be567db00e29ba041ba55be3678c43488a11a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0