• ELISA Kits ›
  • RAGE ELISA Kits
Invitrogen
Rat RAGE (AGER) ELISA Kit
View all (2) RAGE ELISA Kits
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Rat RAGE (AGER) ELISA Kit
Group 53 Created with Sketch.
Rat RAGE (AGER) ELISA Kit
Group 53 Created with Sketch.
FIGURE: 1 / 3
{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}
Rat RAGE (AGER) ELISA Kit
ELISA Standard curve of RAGE (AGER) using Rat RAGE (AGER) ELISA Kit (Product # ERAGER). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product
{{$ctrl.videoDesc}}
{{$ctrl.videoLongDesc}}
Rat RAGE (AGER) ELISA Kit
Rat RAGE (AGER) ELISA Kit (ERAGER)
videoThumbnail
►
Product Specifications
Analytical sensitivity
300 pg/mL
Assay range
0.41-300 ng/mL
Sample type/volume
Plasma
50 µL
Serum
50 µL
Supernatant
100 µL
Hands-on time
1 hr 20 min
Time-to-result
4 hr 45 min
Homogenous (no wash)
No
Interassay CV
<12%
Intraassay CV
<10%
Instrument
Absorbance Microplate Reader
Product size
96 Tests
Contents
Pre-coated 96 well plate
Standard
Assay Diluent concentrate
Biotinylated Detection Antibody
SAV-HRP
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers
Shipping conditions
Wet ice
Storage
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Protein name
RAGE
Protein family
Other Proteins
Species (tested)
Rat
Assay kit format
Sandwich ELISA Kit
Detector antibody conjugate
Biotin
Label or dye
HRP
About This Kit
The Rat RAGE (AGER) ELISA quantitates Rt RAGE in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt RAGE.
Principle of the method
The Rat RAGE solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Target information
Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
References (0)
Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product
Bioinformatics
Gene aliases : RAGE
Gene ID : (Rat) 81722
Gene symbol : Ager
Protein Aliases : advanced glycosylation end product-specific receptor, advanced glycosylation end product-specific receptor variant 2, advanced glycosylation end product-specific receptor variant 3, advanced glycosylation end product-specific receptor variant 4, advanced glycosylation end product-specific receptor variant 5, RAGE-4 ORF3, sRAGE, receptor for advanced glycosylation end products
Recommended Products
IL-6 Human Matched Antibody Pair
Because you are interested in IL-6 Human ELISA Kit.
SIZE
10 x 96 tests
PRICE
USD 400.00
Cat # CHC1263
guarantee_icon
Performance Guarantee
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
help_icon
We're here to help
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support
Your items have has been added!
Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0