• ELISA Kits ›
  • CD146 ELISA Kits

Invitrogen

Mouse MCAM/CD146 ELISA Kit

View all (2) CD146 ELISA Kits

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Technical guide & protocol Tech Support
Mouse MCAM/CD146 ELISA Kit
Group 53 Created with Sketch.
Mouse MCAM/CD146 ELISA Kit
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Mouse MCAM/CD146 ELISA Kit

ELISA Standard curve of MCAM/CD146 using Mouse MCAM/CD146 ELISA Kit (Product # EM52RB). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Mouse MCAM/CD146 ELISA Kit
Mouse MCAM/CD146 ELISA Kit (EM52RB)
videoThumbnail
►

Product Specifications

Analytical sensitivity

0.18 ng/mL

Assay range

0.18-40 ng/mL

Sample type/volume

Serum
50 µL
Plasma
50 µL
Supernatant
50 µL

Hands-on time

1 hr 20 min

Time-to-result

4 hr 45 min

Homogenous (no wash)

No

Interassay CV

<12%

Intraassay CV

<10%

Instrument

Absorbance Microplate Reader

Product size

96 Tests

Contents

Pre-coated 96 well plate
Standard
Assay Diluent concentrate
Biotinylated Detection Antibody
SAV-HRP
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers

Shipping conditions

Wet ice

Storage

Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C

Protein name

CD146

Species (tested)

Mouse

Assay kit format

Sandwich ELISA Kit

Detector antibody conjugate

Biotin

Label or dye

HRP

About This Kit

Mouse MCAM/CD146 quantitates mouse MCAM/CD146 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse MCAM/CD146.

Target information

CD146, also known as MCAM (Melanoma Cell Adhesion Molecule), is an integral membrane glycoprotein belonging to the immunoglobulin superfamily. It is heavily glycosylated, with more than 50% of its mass derived from carbohydrates. CD146 is primarily expressed on endothelial cells at cell-cell junctions, playing a crucial role in cell adhesion and the cohesion of the endothelial monolayer in vascular tissue. In addition to endothelial cells, CD146 is expressed on bone marrow fibroblasts, mesenchymal stromal cells, endometrial stromal cells, and some tumors, particularly melanoma. It has also been observed on a subset of circulating T cells and activated lymphocytes, but not on circulating endothelial cells, as indicated by the absence of other endothelial markers such as CD31 and CD51/61. CD146 functions as a calcium-independent cell adhesion molecule and may also act as a signal transduction molecule. It is involved in the recruitment of Fyn kinase and the subsequent tyrosine phosphorylation of intracellular proteins, which are important for actin cytoskeleton assembly. This signaling role includes triggering tyrosine phosphorylation of FYN and PTK2 and causing a transient increase in intracellular calcium concentration. In the context of cancer, CD146 expression may facilitate the interaction of melanoma cells with the vascular system, enhancing hematogenous tumor spread. Additionally, CD146 could serve as an adhesion molecule active in neural crest cells during embryonic development, highlighting its diverse roles in both normal physiological processes and disease states.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Gene aliases : 1-gicerin, AV025631, CD146, CD149, Mcam, Muc18, s-endo, s-gicerin

Gene ID : (Mouse) 84004

Gene symbol : Mcam

Protein Aliases : CD146, Cell surface glycoprotein MUC18, Gicerin, l-gicerin protein, melanoma adhesion molecule, Melanoma cell adhesion molecule, Melanoma-associated antigen MUC18

UniProt ID (Mouse) Q8R2Y2

Recommended Products

IL-6 Human Matched Antibody Pair

Because you are interested in IL-6 Human ELISA Kit.

SIZE

10 x 96 tests

PRICE

USD 400.00

Cat # CHC1263

guarantee_icon

Performance Guarantee

If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0